missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Micro BCA™ Reagent B (MB)

Catalog No. pi23232
Change view
Click to view available options
Quantity:
240 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI23232 240 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI23232 Supplier Thermo Scientific™ Supplier No. LSG23232
Only null left

Special 3-component version of our popular BCA Reagents to measure total protein concentration of dilute protein solutions (0.5 to 20 micrograms/mL).

Thermo Scientific™ Pierce Micro BCA Protein Assay uses a concentrated form of the BCA Reagents to accurately measure dilute protein solutions (0.5 to 20μg/mL).

Additional Information and Online Ordering

The Micro BCA Protein Assay Kit is a specialized version of the popular Pierce BCA Protein Assay for determining the protein concentration of dilute samples. Mixing together the three Micro BCA Reagents results in a working solution that is sufficiently concentrated to measure protein when mixed with an equal volume of sample. The result is an assay for accurately measuring 0.5 to 20μg/mL protein solutions. The assay is exceptionally linear and exhibits very low levels of protein-to-protein variability.

Highlights:

  • Sensitive – accurately detect down to 0.5μg/mL (2μg/mL in microplate format)

  • High linearity – linear working range for BSA equals 0.5 to 20μg/mL

  • Colorimetric – measure with a standard spectrophotometer or plate reader (562nm)

  • Compatible – unaffected by typical concentrations of most ionic and nonionic detergents

  • Convenient – microplate and cuvette protocols provided with the instructions

  • Stable – kit stable at room temperature; prepared working reagent stable for to 24 hours

Includes:

Three Micro BCA Reagents (MA, MB, MC) and 10 ampules of Albumin Standard (2mg/mL)

Specifications

Content And Storage Upon receipt store product at room temperature. Product shipped at ambient temperature.

Note: If Reagent MB precipitates upon shipping in cold weather or during long-term storage, dissolve precipitates by gently warming and stirring the solution. Discard any reagent that shows discoloration or evidence of microbial contamination.
Description Pierce Micro BCA Reagent B (MB)
Assay BCA Assay
Detection Method Colorimetric
Product Line Micro BCA, Pierce
Product Type Protein Quantitation Assay
Specificity Not Target-Specific
Sufficient For 3200 Microplate Assays
For Use With (Application) Solution-based Detection, Absorbance
For Use With (Equipment) Spectrophotometer, Microplate Reader
Quantity 240 mL
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.