missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Luciferase Cell Lysis Buffer (2X)

Catalog No. PI16189
Change view
Click to view available options
Quantity:
250 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI16189 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI16189 Supplier Thermo Scientific™ Supplier No. LSG16189
  • ONLINE EXCLUSIVE PRICE
    $922.32
    $460.84 / Each

    Save $461.48

    50.0% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Lyse adherent cells or pelleted cell suspensions for analysis of promoter expression in luciferase assays with this optimized protein extraction reagent.

Thermo Scientific™ Pierce Luciferase Cell Lysis Buffer effectively lyses adherent cells or pelleted suspension cells for analysis of promoter expression in luciferase assays.

Pierce Luciferase Cell Lysis Buffer is effective for preparing high-yield mammalian cell lysates from either adherent cells or pelleted suspension cells for analysis in luciferase reporter assays. The reagent is compatible with all Thermo Scientific Pierce Luciferase Assay Kits. The cell lysis buffer has been validated for use with COS-7, NIH3T3, Jurkat and other cell types. Lysates are produced rapidly and easily for luciferase assays and are also compatible with BCA and Coomassie (Bradford) protein assays.

Highlights:

  • Compatible with all Pierce Luciferase Reporter Assays
  • Enables rapid and efficient lysis
  • Tested compatibility with multiple Mammalian cell lines
  • Compatible with protein assays (e.g., Coomassie Plus and BCA)

Specifications

Type Cell Lysis Buffer
Quantity 250 mL
Content And Storage Store at room temperature.
Format Liquid
Do you still offer quantitative IP (qIP) Luciferase assay kits?

Our qIP Luciferase assay kits have been discontinued but we do carry epitope-tagged (HA or c-Myc) and Tluc (TurboLuc luciferase)-tagged mammalian expression vectors, and qIP protein interaction assay reagents for these assay kits.

For Research Use Only. Not for use in diagnostic procedures.