missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ IP Lysis Buffer

Catalog No. p-4531876
Change view
Click to view available options
Quantity:
100 mL
250 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI87787 100 mL
PI87788 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PI87787 Supplier Thermo Scientific™ Supplier No. 87787
Only null left

Maximize cell lysate yield, purity and compatibility with this immunoprecipitation-optimized protein extraction reagent.

Thermo Scientific™ Pierce IP Lysis Buffer is optimized for cell lysate yield, purity and compatibility with immunoprecipitation.

The IP Lysis Buffer is a mammalian whole cell lysis reagent based on a modified RIPA buffer formulation without SDS. This moderate-strength lysis buffer effectively solubilizes cellular proteins but does not liberate genomic DNA or disrupt protein complexes like ordinary RIPA buffer. Pierce IP Lysis Buffer is specially formulated for pulldown and immunoprecipitation assays.

Highlights:

  • Optimized for compatibility with immunoprecipitation and pull-down assays
  • Compatible with protein assays, reporter assays and immunoassay procedures
  • Ready-made formula is effective for extracting cytoplasmic, membrane and nuclear proteins
  • Gentle formulation helps maintain protein complexes for co-immunoprecipitation (co-IP)
  • Does not liberate genomic DNA which can cause high sample viscosity

Specifications

Quantity 100 mL
Format Liquid
Product Type IP Lysis Buffer
Volume (Metric) 100 mL
Content And Storage Upon receipt store at 4°C.
Product Line Pierce

For Research Use Only. Not for use in diagnostic procedures.