missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Mannan Binding Protein Agarose

Catalog No. PI22212
Change view
Click to view available options
Quantity:
10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI22212 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI22212 Supplier Thermo Scientific™ Supplier No. LSG22212

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Easily purify IgM from mouse ascites fluid using immobilized mannan binding protein, which binds to IgM in the kit-supplied buffers.

Thermo Scientific™ Pierce IgM Purification Kit is an easy-to-use affinity matrix and buffer system to purify IgM from mouse ascites fluid; the method uses immobilized mannan binding protein, which binds to IgM in the kit-supplied buffers.

Pierce MBP Agarose is purified mannan binding protein (also called mannose-binding lectin, MBL) that has been immobilized on 4% beaded agarose. When used with an optimized buffer system based on temperature- and calcium-dependent binding, this resin enables specific affinity purification of mouse IgM from ascites fluid and other concentrated samples. The resin and essential buffers are available individually or bundled together in a complete IgM Purification Kit containing a pre-packed 5mL column of the MBP Agarose.

Highlights:

  • Immobilized MBP – affinity system based on mannan binding protein (MBP) allows specific purification of mouse IgM from ascites fluid or partial isolation of human IgM from serum

  • Complete kit – IgM Purification Kit includes a pre-packed MBP column and ready-to-use buffers for convenient purification of at least 1.5mg IgM from 0.5mL ascites samples per column-use

  • Mild conditions – nondenaturing, near-neutral buffer conditions ensure recovery of functional antibodies

  • Reusable – stable ligand and resins can be regenerated and reused at least 10 times without significant decline in binding capacity

Includes:

Kit contains a 5mL MBP Agarose Column, column preparation buffer, IgM binding and elution buffers, and a column extender

Specifications

Product Type Agarose
Product Line Pierce
Column Type Agarose Resin, Affinity
Format Bottle
Stationary Phase Lectin
Purification Target IgM Antibody
Description Pierce Mannan Binding Protein Agarose
Content And Storage Upon receipt store product at 4°C. Product is shipped at ambient temperature.
Quantity 10 mL

For Research Use Only. Not for use in diagnostic procedures.