missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Hydrochloric Acid Solution, Sequencing Grade

Catalog No. PI24308
Change view
Click to view available options
Quantity:
10 x 1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI24308 10 x 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI24308 Supplier Thermo Scientific™ Supplier No. LSG24308

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Constant-boiling 6N HCl solution, supplied in 1mL ampules, used to acid-hydrolyze proteins (protein hydrolysis) for HPLC amino acid composition analysis.

Thermo Scientific™ Pierce Hydrochloric Acid Solution is a constant-boiling 6N HCl solution, supplied in 1mL ampules, that is specifically formulated as a sequencing-grade reagent for total protein hydrolysis for amino acid analysis.

Highlights:

  • Hydrolyzes peptides in 6 hours at 150°C
  • Specially purified to give ninhydrin-negative blank on hydrolysis
  • Packaged in prescored ampules to eliminate contamination and ensure product integrity

Ordering Information:

X in the ordering table indicates that hazardous shipping charges apply.

Specifications

Product Type Hydrochloric Acid Solution
Content And Storage No product PDF available for storage information.
Quantity 10 x 1 mL
Form Solid

For Research Use Only. Not for use in diagnostic procedures.