missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Protein G Plus UltraLink™ Resin

Extremely durable polyacrylamide support with densely immobilized, recombinant Protein G for various IgG purification methods.

Supplier:  Thermo Scientific™ 53128

Catalog No. PI53128

Only null left
Explore available promotions

Description

Description

Thermo Scientific™ Pierce Protein G Plus UltraLink Resin is an extremely durable beaded polyacrylamide support to which recombinant Protein G has been immobilized at high density for use in IgG purification methods.

The Protein G Plus UltraLink Resin is used for high-capacity binding and purification of IgG in gravity-flow, spin-column or medium-pressure chromatography methods. Protein G is a popular antibody-binding protein that is especially useful for purification of IgG from mouse, rat and human serum.

UltraLink Resin is an azlactone-activated polyacrylamide support that is hydrophilic, charge-free, high capacity, highly crosslinked, rigid and porous. This support is especially useful for medium pressure techniques when using large sample volumes requiring fast-flow techniques (FPLC) and large-scale applications. Unlike typical beaded agarose supports, UltraLink Support does not crush or collapse when subjected to high flow rates that result in pressures exceeding 25psi.

Highlights:

  • Immobilized Protein G – specifically binds IgG from several mammalian species, especially all isotypes of IgG from human, mouse and rat

  • High capacity – “Plus” variety has a dense load of immobilized Protein G, providing a binding capacity of at least 25mg human IgG per milliliter of resin

  • UltraLink Resin – acrylamide-based support provides an alternative to crosslinked beaded agarose supports for medium pressure applications

Properties of Thermo Scientific UltraLink Resin:

  • Support pH Stability: 1 to 13

  • Average Particle Size: 50 to 80 microns

  • Exclusion Limit: greater than 2,000,000 daltons

  • Average Surface Area: greater than 250m2 per gram of beads

  • Average Pore Volume: greater than 1.2mL per gram of beads (> 60% of bead volume)

  • Pore Size: 500 to 1000 angstroms (50 to 100nm)

  • Maximum Linear Velocity: 3000cm per hour

  • Maximum Pressure: 100psi (6.9 bar), defined as the maximum pressure drop across a column that the resin can withstand (Note: The indicated gauge pressure of a liquid chromatography apparatus may be measuring the total system pressure rather than the pressure drop across the column.)
Specifications

Specifications

Resin
UltraLink Affinity Resin
Protein G
Upon receipt store product at 4°C. Products are shipped at ambient temperature.
Pierce, UltraLink
Bottle
IgG Antibodies
2 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

For Research Use Only. Not for use in diagnostic procedures.