missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ HABA (4'-hydroxyazobenzene-2-carboxylic acid)

Catalog No. pi28010
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI28010 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI28010 Supplier Thermo Scientific™ Supplier No. 28010

A colored, biotin assay reagent HABA (4'-hydroxyazobenzene-2-carboxylic acid) to measure biotinylation levels of biotinylated antibodies or proteins.

Thermo Scientific™ Pierce HABA is 4'-hydroxyazobenzene-2-carboxylic acid, a simple reagent that enables spectrophotometric (colorimetric) estimation of biotinylation levels of labeled proteins and other molecules.

Thermo Scientific Pierce HABA is 4'-hydroxyazobenzene-2-carboxylic acid, a simple reagent that enables spectrophotometric (colorimetric) estimation of biotinylation levels of labeled proteins and other molecules. Determine the molar ratio of biotin incorporated into a protein using the HABA-Avidin method. The HABA dye (4'-hydroxyazobenzene-2-carboxylic acid ) binds to avidin to produce a yellow-orange colored complex which absorbs at 500nm. Free biotin will displace the HABA dye and cause the absorbance to decrease. A standard curve can be established using the free biotin to estimate the number of moles of biotin incorporated after biotinylating a protein.

Highlights:

  • HABA-avidin complex can be used over a wide range of pH and salt concentrations
  • Amount of Avidin can be calculated directly from the increased absorbance at 500nm complexing with the HABA Dye
  • Calculate results directly from absorbance values based on extinction coefficients using the procedure outlined in the instructions or use Pierce's online HABA calculator
  • Complete kits also available! See Pierce Biotin Quantitation Kit (Part No. 28005) and Fluorescence Biotin Quantitation Kit (Part No. 46610)

Specifications

Product Type HABA
Quantity 10 g
Content And Storage Upon receipt store product at room temperature. Product shipped at ambient temperature.
Product Line Pierce
Label or Dye Biotin
Labeling Target Antibodies, Proteins
How can I determine the amount of biotinylation on my ligand?

An assay with HABA dye (Cat. No. 28010 or 28005) can be used to determine the molarity of biotin in solution.

For Research Use Only. Not for use in diagnostic procedures.