missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ GST Agarose

Catalog No. PI20211
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI20211 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI20211 Supplier Thermo Scientific™ Supplier No. LSG20211
Season of Savings
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Isolate or remove cross-reacting GST-specific serum or other antibody solutions with immobilized glutathione S-transferase agarose.

Perform isolation or removal of cross-reacting GST-specific antibodies from serum or other antibody solutions with Thermo Scientific™ Pierce GST Agarose.

This GST resin consists of purified glutathione S-transferase protein from Schistosoma japonicum that has been immobilized on crosslinked 6% beaded agarose. Antibody molecules in that cross-react with the GST portion of a recombinant fusion protein can be removed from the desired protein-specific antibody by passing antiserum over a GST agarose column. GST antibodies will bind to the immobilized GST, whereas antibodies against other fusion proteins will pass through the column unrestrained.

Highlights:

  • Specific – provides purification of target antibodies from those directed against the GST tag

  • Reusable – bound antibodies can be eluted with 6M guanidinium chloride allowing for 10 purifications with each column or aliquot of resin

  • Two formats – choose either pre-packed columns or resin slurry for use at a variety of scales

Specifications

Product Type Agarose
Product Line Pierce
Column Type Agarose Resin, Affinity
Format Bottle
Stationary Phase GST Protein
Purification Target Anti-GST Antibody
Description Pierce GST Agarose
Content And Storage 4°C
Quantity 5 mL

For Research Use Only. Not for use in diagnostic procedures.