missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Gentle Ag/Ab Elution Buffer, pH 6.6

Catalog No. PI21027
Change view
Click to view available options
Quantity:
500 mL
3.75 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI21027 500 mL
PI21013 3.75 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PI21027 Supplier Thermo Scientific™ Supplier No. 21027
Season of Savings
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Ensure high-yield, nondenaturing antibody purification using IgG affinity purification supports with these gentle buffer solutions.

Thermo Scientific™ Pierce IgG Binding and Elution Buffers ensure high-yield, nondenaturing antibody purification with IgG affinity purification supports, such as Protein A and Protein G agarose beads.

These products include IgG Binding Buffers specific for each immunoglobulin-binding protein (Protein A, Protein G, Protein A/G). The Pierce IgG Elution Buffer provides efficient dissociation and elution of antibodies from all three IgG-binding proteins, including Protein L. This low-pH elution buffer is also effective for most antibody-antigen or protein-protein affinity purification systems.

The Gentle Ag/Ab Binding and Elution Buffer system uses near-neutral, high-salt conditions for elution, enabling even sensitive and labile antibodies (or other proteins) to be eluted from an affinity system without denaturation and inactivation. The phosphate-free binding buffer in this system is a necessary substitute for the Protein A binding buffer (pH 8), which precipitates with the Gentle Ag/Ab Elution Buffer.

Finally, the Mouse IgG1 Mild Elution Buffer, when used in combination with Protein A resin, enables selective elution and separation of the IgG1 subclass from total bound mouse IgG.

Highlights:

  • Three optimized binding buffers – for Protein A, Protein G or Protein A/G

  • Two elution buffer options – acidic (pH 2.8) or high salt (near-neutral)

  • Special system for mouse IgG1 – selective, two-stage elution from Protein A agarose

Item-Specific Features:

  • Protein A IgG Binding Buffer: pH 8.0; phosphate-based; contains glycerol and EDTA

  • Protein A/G IgG Binding Buffer: pH 8.0; phosphate-based; contains EDTA

  • Protein G IgG Binding Buffer: pH 5.0; contains 0.02% sodium azide

  • IgG Elution Buffer: pH 2.8; amine-based

  • Gentle Ag/Ab Binding Buffer: pH 8.0; phosphate-free; contains 0.05% sodium azide

  • Gentle Ag/Ab Elution Buffer: pH 6.6; high salt (ionic strength)

  • Mouse IgG1 Mild Elution Buffer: pH 6.0; contains 0.02% sodium azide

Includes:

Kits contain both binding and elution buffers

Specifications

Chemical Name or Material Elution Buffer
pH 6.6
Physical Form Liquid
Product Line Pierce
Quantity 500 mL
After immobilizing my antibody, the first affinity purification of my protein worked well, but the protein failed to bind to the antibody during the second round of purification. What did I do wrong?

Low pH elution is the most commonly used method of elution for affinity purification, however, it could lead to denaturation of some antibodies which will negatively affect subsequent antigen binding. To prevent this from happening, after protein purification and column regeneration, immediately wash the column with neutralization or binding buffer and store at 4 degrees C in buffer containing an antimicrobial agent such as sodium azide.

Alternatively, use a near-neutral, high salt elution buffer such as Pierce Gentle Ag/Ab Elution Buffer (Cat. No. 21013, 21027, or 21030). For alternative elution buffers, please refer to this Tech Tip (https://tools.thermofisher.com/content/sfs/brochures/TR0027-Elution-conditions.pdf).

How many purifications can be performed using the same affinity column?

This varies with the stability of the immobilized protein and the type of elution buffer used. Columns can typically be reused at least 10 times without significant loss of activity. If the immobilized molecule is sensitive to low pH, consider using our Pierce Gentle Ag/Ab Elution Buffer (Cat. No. 21013, 21027 or 21030). For alternative elution buffers, please refer to this Tech Tip(http://tools.thermofisher.com/content/sfs/brochures/TR0027-Elution-conditions.pdf).

For Research Use Only. Not for use in diagnostic procedures.