Learn More
Thermo Scientific™ Pierce™ Gaussia Luciferase Flash Assay Kit
Assay regulatory element transcriptional activity in mammalian culture media or whole cell lysates with this highly sensitive flash-based Gaussia luciferase kit.
Supplier: Thermo Scientific™ 16159
Description
The Thermo Scientific™ Pierce Gaussia Luciferase Flash Assay Kit provides researchers with the most sensitive assay for transcriptional activity of regulatory elements in mammalian cell culture media and whole cell lysate.
This Flash Assay Kit contains reagents for measuring the activity of Gaussia luciferase in mammalian cell culture media and lysates. When used with Thermo Scientific Gaussia Luc Vectors, the kit provides an extremely sensitive bioluminescent reporter assay system for secreted or intracellular detection of promoter or pathway activity. Gaussia luciferase is highly secreted into the cell culture media, allowing for live cell monitoring of reporter activity. The signal produced by Gaussia is considerably greater than signal from either firefly or Renilla luciferases assayed under similar conditions.
Highlights:
Sensitive – 1000 times greater sensitivity allows utilization of smaller numbers of cells
Cost effective – highest sensitivity assays result in decreased reagent consumption
Secreted luciferase – allows real-time assays and kinetic studies without destroying cells
Time-saving – assays using secreted luciferases require minimal sample handling
Automation-friendly – amenable to high throughput screens
Convenient – contains a universal cell lysis buffer and optimized flash assay reagent
Safe – allows one to perform non-radioactive assays
Includes:
Cell lysis buffer, reaction buffer and substrate
Requires:
Gaussia luciferase and luminometer or other instrument capable of monitoring luminescence, such as Thermo Scientific Luminoskan Ascent and Varioskan Flash Microplate Readers
Recommended for:
Promoter studies for analyzing cis- regulatory elements and trans-acting factors; Drug screening; siRNA and miRNA screening; Multiplexed assays to study off-target effects; Secretory pathway / protein localization reporter assays; Signal transduction pathway analysis; RNA splicing studies
Specifications
| Assay Kit | |
| 1000 Luciferase Assays | |
| Sufficient For: 1000 luciferase assays in microplate wells • Gaussia Flash Assay Buffer, 50 mL (store at 4°C) • 100X Coelenterazine, 0.5 mL (store at -80°C) • Luciferase Cell Lysis Buffer (2X), 60 mL (store at room temperature) Kit may be stored at -80°C, or individually at indicated temperatures. |
|
| Mammalian Cells | |
| Luminometer (Microplate) | |
| Enzyme Labeled | |
| Enhanced Chemiluminescence | |
| Pierce |
| Luciferase, Gaussia Luciferase | |
| 1000 Reaction Kit | |
| Reporter Enzyme, Luciferase Reporter Assay | |
| Bioluminescence | |
| 384-well plate, 96-well plate | |
| Chemical Substrate | |
| Luciferase Substrate | |
| Coelenterazine |
For Research Use Only. Not for use in diagnostic procedures.