Learn More
Thermo Scientific™ Pierce™ Gaussia-Firefly Luciferase Dual Assay Kit
Description
Simultaneously detect intracellular Gaussia and Red Firefly luciferase activity in mammalian whole cell lysates.
The Thermo Scientific™ Pierce Gaussia-Firefly Luciferase Dual Assay Kit provides the necessary reagents to simultaneously detect intracellular Gaussia and Red Firefly luciferase activity in mammalian whole cell lysates.
The Pierce Gaussia-Firefly Luciferase Dual Assay Kit is a highly sensitive assay that allows for the simultaneous detection of Gaussia and Red Firefly luciferase activity. Gaussia luciferase acts as an experimental reporter with constitutively active red firefly as a normalization control. This reporter-and-control combination enables simultaneous monitoring of experimental reporter and control luciferase activities in a single-read assay without the need for two-step addition of substrate reagents or quenching.
The assay working solution contains substrates for both luciferases, and the reactions occur simultaneously with flash-type kinetics. The resulting luminescent signals are spectrally resolvable using filters. In a single sample, researchers can assay transcriptional activity of regulatory elements, signal transduction pathways, and effects of activators or inhibitors.
Highlights
- Simultaneous—concurrent, filter-based, wavelength-separated detection of two luciferase activities
- Sensitive—measure bright blue Gaussia and red firefly luciferase activities in the same sample
- Fast—no quenching step required, unlike in traditional sequential dual assays
- Multiplex—capable of quantitating two cellular activities in the same sample(s)
Requires
Gaussia and Red Firefly luciferase reporters; filter sets in the ranges of 425 to 525 nm for Gaussia Luc and 615 to 675 nm for Red Firefly Luc; luminometer or other instrument capable of monitoring luminescence, such as the Varioskan ALF or the Varioskan LUX microplate readers; injector for assessing more than 24 wells at a time
Recommended applications
Study two regulatory elements at the same time; monitor two signaling pathways simultaneously; enable studying more than one target per screen (e.g., off-target effects)
Specifications
Specifications
| Type | Assay Kit |
| Target | Luciferase, Firefly Luciferase, Gaussia Luciferase |
| Quantity | 100 Reaction Kit |
| Content And Storage | • Gaussia-Firefly Dual Assay Buffer, 5 mL, store at 4°C • Coelenterazine (100X), 50 μL, store at -80°C • D-Luciferin, Lyophilized, 3 mg, store at 4°C • 2X Cell Lysis Buffer, 6 mL, store at room temperature Upon receipt store kit at -80°C or store individual components as indicated above. |
| Assay | Reporter Enzyme, Luciferase Reporter Assay |
| Compatible Cells | Mammalian Cells |
| Detection Method | Bioluminescence |
| For Use With (Equipment) | Luminometer (Microplate) |
| Format | 384-well plate, 96-well plate |
| Label Type | Enzyme Labeled |
| Show More |
Frequently Asked Questions (FAQs)
Here are possible causes and solutions:
- Non-specific oxidation of substrate: Use a new sample; Avoid repeated freezing and thawing of the sample.
- Control sample is contaminated: Change pipette tips after each well; Reduce shaker speed during the cell lysis step to avoid contaminating the wells.
This could be due to high luciferase expression. Here are some suggestions:
Note: A low sample volume can increase assay variability. Dilute the sample and use the recommended volume of 10-20 µL per assay.
Here are possible causes and solutions:
- Low luciferase expression: Lyse cells in smaller volume of 1X Cell Lysis Buffer; Use a different promoter or growth conditions to improve expression; Increase the integration time on the instrument; Scale up volume of sample and reagent per well.
Here are possible causes and solutions:
- Low transfection efficiency: Optimize transfection conditions using a visual transfection control (e.g., a plasmid over-expressing a fluorescent protein); Verify plasmid DNA quality using restriction digestion and agarose gel electrophoresis; Use only transfection grade DNA - please note that most high-quality plasmid DNA should be supercoiled; Use actively dividing, low-passage cells; Use a different cell type.
- No promoter induction: Incubate cells using promoter-specific inducing conditions; Incubate cells for a longer time after treatment; Change growth conditions to improve expression; Use a different promoter.
- Substrate auto-oxidized: Protect substrate from light and air; Maintain 100X Coelenterazine at -80 degrees C, 100X Vargulin at -20 degrees C, and 100X D-Luciferin at -20 degrees C.
Here are possible causes and solutions:
- Non-specific oxidation of substrate: Use less serum in the cell culture media; Note: Albumin can increase the auto-oxidation of Vargulin; Avoid repeated freezing and thawing of the sample.
- Control sample is contaminated: Use new sample; Change pipette tips after each well.
For Research Use Only. Not for use in diagnostic procedures.