missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Fluoraldehyde™ o-Phthaldialdehyde Reagent Solution (OPA)

Catalog No. PI26025
Change view
Click to view available options
Quantity:
945 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI26025 945 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI26025 Supplier Thermo Scientific™ Supplier No. 26025

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Ready-to-use solution of o-phthalaldehyde, a fluorescent, amine-reactive, HPLC pre- or post-column labeling reagent to measure peptides or amino acids (Ex 340nm, Em 455nm).

Thermo Scientific™ Pierce Fluoraldehyde (OPA) Reagent is a ready-to-use solution of o-phthalaldehyde, a fluorescent pre- or post-column derivatization reagent for peptide or amino acid detection and quantitation in HPLC.

Fluoraldehyde Reagent contains OPA (o-phthalaldehyde), which reacts with primary amines of amino acids, peptide and proteins to enable fluorescent detection and quantitation. The reagent that can be used as a protein or peptide assay reagent or as a pre- or post-column detection reagent for amino acid analysis (HPLC). Reaction of OPA with proteins and peptides yields linear results over a wide range of concentrations. The Fluoraldehyde Reagent Solution is supplied ready to use and enables fast quantitation of proteins or peptides in solution.

Highlights:

  • For pre- or post-column amino acid derivatization for fluorescent detection and quantitation

  • Reacts with all primary amine-containing analytes to yield fluorescent isoindole derivatives

  • Provides an accurate measure of both composition and absolute protein-peptide content

  • Ideal for work with recombinant proteins and synthetic peptides

  • Can be used for fluorescent protein or peptide assay

  • Pre-column derivatization mixtures can be injected into LC without any processing

Reagent Properties:

  • Formulation: 0.8mg/mL o-phthalaldehyde (OPA) in proprietary buffer, pH 10

  • Form: Clear, colorless to slightly yellow liquid

  • Reactive toward: Primary amines (–NH2)

  • Detection: Fluorescence (excitation/emission = 340nm/455nm)

  • Application: Pre- or post-column derivatization for detection by HPLC

Specifications

Product Type o-Phthaldialdehyde
Chemical Reactivity Amine
Product Line Fluoraldehyde
Content And Storage Store under nitrogen at 4°C away from oxidizing reagents and direct sunlight.
Label or Dye OPA (o-phthaldialdehyde)
Detection Method Fluorescence
Quantity 945 mL
Form Liquid
What is the shelf life for Fluoraldehyde o-Phthaldialdehyde Reagent Solution (OPA) (Cat. No. 26025)?

The lot-specific expiration date for Fluoraldehyde o-Phthaldialdehyde Reagent Solution (OPA) (Cat. No. 26025) can be found on the Certificate of Analysis.

For Research Use Only. Not for use in diagnostic procedures.