missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ FITC Antibody Labeling Kit

Catalog No. PI53027
Change view
Click to view available options
Quantity:
3 reactions kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI53027 3 reactions kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI53027 Supplier Thermo Scientific™ Supplier No. 53027

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Activated derivatives of green-fluorescent fluorescein dye to label antibodies, proteins and other molecules for fluorescence imaging (ex 494; em 518).

Thermo Scientific™ FITC and Fluorescein Labeling Reagents and Kits are high-performance derivatives of fluorescein dye, activated for easy and reliable labeling of antibodies, proteins and other molecules for use as fluorescent probes.

Fluorescein dyes are used in wide-ranging applications including fluorescence microscopy, flow cytometry and immunofluorescence-based assays such as Western blotting and ELISA. FITC and NHS-fluorescein are reactive toward primary amine groups on proteins, peptides and other biomolecules, while fluorescein-5-maleimide and 5-IAF react with free sulfhydryls on cysteine residues.Most activated fluorescein derivatives are mixture of isomers with reactive groups attached at the 5- and 6-positions of the bottom ring. The properties of these isomers are indistinguishable in terms of excitation and emission spectra and for protein applications there is no need to isolate a specific isomer. The single isomer is used in high resolution separation techniques such as sequencing and capillary electrophoresis.

Highlights:
Amine-specific labeling: the NHS-ester or isothiocyanate efficiently labels antibodies and other purified proteins at primary amines (lysine side chains) Optimized procedure: following the standard protocol results in antibodies with excellent dye:protein ratios for optimum activity and fluorescence Single-use fluors: no need to weigh tiny amounts powder; kits contain single-use vials of reagent Efficient purification: kits include purification resin and easy-to-use spin columns, ensuring rapid and efficient removal of non-reacted dye and excellent protein recovery

Specifications

Description Pierce FITC Antibody Labeling Kit
Product Type Labeling Kit
Quantity 3 reactions kit
Sufficient For Labeling and purifying 3 x 1 mg (2 mg/mL) of IgG or other proteins
Content And Storage
  • FITC, 3 x 50 μg vials
  • Borate Buffer, 1 mL
  • Purification Resin, 5 mL
  • Spin Columns, 6 each
  • Collection Tubes, 12 each
Excitation/Emission 494/518
Labeling Scale 0.5 to 1 mg
Product Line Pierce
Solubility Aqueous Buffer
Label or Dye FITC (Fluorescein)
Labeling Target Antibodies, Proteins
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.