Learn More
Thermo Scientific™ Pierce™ Firefly Luciferase Flash Assay Kit
Description
Assay regulatory element transcriptional activity in whole cell lysates with this highly sensitive flash-based firefly luciferase kit.
The Thermo Scientific™ Pierce Firefly Luciferase Flash Assay Kit provides reagents to measure Red firefly luciferase activity in whole cell producing a very strong flash signal which decays rapidly.
Thermo Scientific Pierce Firefly Luciferase Flash Assay Kit contains reagents to measure the activity of firefly luciferase in mammalian whole cell lysates. Containing a convenient lysis buffer, assay buffer and firefly substrate, this kit provides a complete reagent system for the intracellular detection of promoter or pathway activity. Light output resulting from the luciferase reaction is captured using a luminometer and can be correlated with the amount of firefly luciferase protein produced and used to determine the activity of the promoter driving firefly expression. These reagents are optimized to work with Thermo Scientific Firefly Luc Plasmids; however, the Flash Assay Kit may be used to detect the activity of other firefly or ATP-dependent luciferases utilizing D-luciferin as a substrate.
Highlights
- Highly sensitive detection of firefly luciferase activity
- Use with Pierce Firefly Luciferase Signal Enhancer for best results (maximum signal)
- Working solution stable for up to 2 months at -20°C
- Assay reagents compatible with other firefly luciferases
- Amenable to automation
- Contains a universal cell lysis buffer and optimized flash assay reagent
- Safe non-radioactive assay
Requires
Firefly luciferase and luminometer or other instrument capable of monitoring luminescence, such as the Varioskan ALF or the Varioskan LUX microplate readers; For best results (maximum signal), use with Thermo Scientific Pierce Firefly Luciferase Signal Enhancer
Recommended applications
Promoter studies for analyzing cis- regulatory elements and trans-acting factors; Drug screening; siRNA and miRNA screening; Multiplexed assays to study off-target effects; Secretory pathway/protein localization reporter assays; Signal transduction pathway analysis; RNA splicing studies
Specifications
Specifications
| Type | Assay Kit |
| Target | Luciferase, Firefly Luciferase |
| Quantity | 100 Reaction Kit |
| Content And Storage | • Firefly Flash Assay Buffer, 5 mL, store at 4°C • D-Luciferin, Lyophilized, 3 mg, store at 4°C • 2X Cell Lysis Buffer, 6 mL, store at room temperature Upon receipt store kit at 4°C or store individual components as indicated above. |
| Assay | Reporter Enzyme, Luciferase Reporter Assay |
| Compatible Cells | Mammalian Cells |
| Detection Method | Bioluminescence |
| For Use With (Equipment) | Luminometer (Microplate) |
| Format | 384-well plate, 96-well plate |
| Label Type | Enzyme Labeled |
| Show More |
Frequently Asked Questions (FAQs)
This could be due to contamination of the control sample. Make sure to use new sample and change pipette tips after each well.
This could be due to high luciferase expression. Here are some suggestions:
Note: A low sample volume can increase assay variability. Dilute the sample and use the recommended volume of 10-20 µL per assay
Here are possible causes and solutions:
- Low transfection efficiency: Optimize transfection conditions using a visual transfection control (e.g., a plasmid over-expressing a fluorescent protein); Verify plasmid DNA quality - use only transfection grade DNA; Use actively dividing, low passage cells; Use a different cell type.
- No or low promoter activity: Use conditions known for promoter activation; Incubate cells for a longer time; Change growth conditions to improve expression; Use a different promoter.
- D-luciferin auto-oxidized: Protect substrate from light and maintain 100X D-luciferin at -20 degrees C; Prepare new Working Solution if used longer than 4 hours.
- Low luciferase expression: Use Pierce Firefly Signal Enhancer (100X) (Cat. No. 16180); Lyse cells in a smaller volume of 1X Cell Lysis Buffer; Use a different promoter or growth conditions to improve expression; Increase the integration time on the instrument; Scale-up the volume of sample and reagent per well.
- Degraded luciferase protein: Store cell lysates on ice and perform assays immediately following cell lysis.
Here are possible causes and solutions:
- Non-specific oxidation of substrate: Use less serum in the cell culture media; Note: Albumin can increase the auto-oxidation of Vargulin; Avoid repeated freezing and thawing of the sample.
- Control sample is contaminated: Use new sample; Change pipette tips after each well.
This could be due to high luciferase expression. Here are some suggestions:
Note: A low sample volume can increase assay variability. Dilute the sample and use the recommended volume of 10-20 µL per assay.
For Research Use Only. Not for use in diagnostic procedures.