missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ FDAA (Marfey's Reagent) (1-fluoro-2-4-dinitrophenyl-5-L-alanine amide)

Catalog No. PI48895
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI48895 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI48895 Supplier Thermo Scientific™ Supplier No. LSG48895

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Label and measure optical isomers of amino acids by reverse-phase chromatography (HPLC) with this amine-reactive, colored (Amax 340nm) compound.

Thermo Scientific Pierce FDAA (Marfey's Reagents) reacts with primary amines to enable the separation and quantitation of optical isomers of amino acids by reverse-phase chromatography.

Highlights:

  • Optical isomers of amino acids derivatization complete in 90 minutes
  • Derivatives have an absorption coefficient of ∽3 x 104 Derivatives can be detected by UV at 340nm with picomole sensitivity

Specifications

Product Type FDAA
Chemical Reactivity Amine
Content And Storage Upon receipt store product at room temperature.
Label or Dye Other Label or Dye
Quantity 50 mg
Form Solid

For Research Use Only. Not for use in diagnostic procedures.