missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Fast Blocking Buffer

Catalog No. pi37576
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI37576 100 mL
PI37575 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PI37576 Supplier Thermo Scientific™ Supplier No. 37576

Optimize Western blotting systems by decreasing both background signal and incubation time compared to traditional blocking agents with Thermo Scientific™ Pierce Fast Blocking Buffer, a ready-to-use blocker solution.

Pierce Fast Blocking Buffer effectively and reliably blocks western blots in just five minutes to provide low-background results for traditional western blotting protocols.

Pierce Fast Blocking Buffer streamlines western blot protocols by providing consistent, high-quality blocking for nitrocellulose or PVDF membranes in only five minutes, producing results comparable to traditional blockers. Blocking and probing steps with Fast Blocking Buffer prevent nonspecific binding of detection antibodies during probing steps but allows specific detection to occur. Pierce Fast Wash Buffer provides efficient membrane washing after incubation with a primary and secondary antibody to effectively shorten total washing time to 20 minutes (from the typical 90 minutes). This buffer is compatible with antibodies and biotin-avidin systems, as well as both nitrocellulose and PVDF membranes.

Related products

Pierce Fast Wash Buffer, 10X (Cat. No. 37577)

Specifications

Chemical Name or Material Blocking Buffer
Concentration 1X
For Use With (Application) Western Blot
Physical Form Liquid
Quantity 100 mL
How can I reduce background bands in my Western blot?

Optimize the concentration of primary and secondary antibodies. In some cases, increasing the concentration of blocking agent (BSA or non-fat dry milk) or usiing an alternative blocking solution such as Starting Block or SuperBlock may reduce background signal. After incubation with the primary antibody, wash at least 2 times with TBST (include 0.5 M NaCl in one or more of the wash steps). Avoid Nonidet P40 or Triton X-100 in buffers because protein detection is decreased when these detergents are used.

For Research Use Only. Not for use in diagnostic procedures.