missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Fast Wash Buffer, 10X

Catalog No. PI37577
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI37577 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI37577 Supplier Thermo Scientific™ Supplier No. 37577
Season of Savings
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Optimize Western blotting systems by decreasing both background signal and incubation time compared to traditional blocking agents with this ready-to-use blocker solution.

Optimize Western blotting systems by decreasing both background signal and incubation time compared to traditional blocking agents with Thermo Scientific™ Pierce Fast Blocking Buffer, a ready-to-use blocker solution.

Additional Information and Online Ordering

Pierce Fast Blocking Buffer streamlines Western blot protocols by providing consistent, high-quality blocking for nitrocellulose or PVDF membranes in only 5 minutes. Blocking and probing steps with Fast Blocking Buffer prevents nonspecific binding of detection antibodies during probing steps but allows specific detection to occur. Pierce Fast Wash Buffer provides efficient membrane washing after incubation with a primary and secondary antibody to effectively shorten total washing time to 20 minutes (from the typical 90 minutes). These two buffers are compatible with antibodies and biotin-avidin systems, as well as both nitrocellulose and PVDF membranes. The blocking buffer provides rapid and effective blocking in just five minutes while producing results comparable to traditional blockers for Western blots.

Highlights:

  • Fast – use both buffers to shorten the typical Western blot development by over 2 hours

  • Simple – optimized protocol makes Western blot analysis easier than ever

  • Low background – provides results comparable to classic Western blotting buffers

Specifications

Chemical Name or Material Wash Buffer
Description Pierce Fast Wash Buffer, 10X
For Use With (Application) Western Blot
Physical Form Liquid
Product Line Pierce
Quantity 1 L

For Research Use Only. Not for use in diagnostic procedures.