missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Fab Preparation Kit

Catalog No. PI44985
Change view
Click to view available options
Quantity:
10 Antibody Samples, each containing 0.25 to 4 mg IgG kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI44985 10 Antibody Samples, each containing 0.25 to 4 mg IgG kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI44985 Supplier Thermo Scientific™ Supplier No. LSG44985

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Digest human or mouse IgG antibodies with immobilized papain protease and purify Fab fragments using Protein A agarose.

Thermo Scientific Pierce Fab Preparation Kit uses immobilized papain protease to digest human or mouse IgG antibodies to make separate Fab and Fc fragments and subsequently to purify the Fab using Protein A agarose.

Features of the Fab Preparation Kit:

  • High capacity—use the standard size kit for fragmenting 0.25 to 4 mg IgG
  • Enzyme-free digestion products—Immobilized Papain (beaded agarose resin) provides for control of the digestion reaction and complete removal of resulting antibody fragments from the proteolytic enzyme
  • Suitable for human and other species of IgG—the kit procedure is optimized for human, mouse and rabbit IgG. Papain-based digestion is effective for certain other species and subclasses of IgG (e.g., goat and pig), although the purification step requires that the antibody effectively binds to Protein A. (Note: for best results with mouse IgG1, use Part No. 44980.)
  • Provides ready-to-use Fab—digestion and final recovery of purified Fab fragments occurs in neutral pH sodium phosphate buffer, suitable for storage or immediate use in typical applications
  • Complete—kits include all reagents needed to prepare and purify antibody fragments
  • Fast—spin format greatly reduces sample processing time
  • Flexible—Protocols are included for multiple species and IgG subclasses, as well as sample size and concentration
  • Efficient—enhanced yield and sample purity

These Fab Preparation Kits are suitable for human, rabbit, mouse and other species and subclasses of IgG. The papain antibody digestion reaction is performed in convenient disposable spin columns that allow efficient removal of the immobilized protease and maximum recovery of the IgG fragments. Also included in the kits are Thermo Scientific NAb Protein A Spin Columns and buffers to efficiently purify the resulting fragments. Protein A binds the Fc fragments and undigested IgG, allowing the pure Fab fragments to be recovered in the flow-through fraction. The kits also include Thermo Scientific Zeba Desalting Spin Columns for preparing the IgG sample quickly without dilution instead of utilizing time-consuming dialysis steps.

The kits use papain, a nonspecific thiol-endopeptidase, to enzymatically cleave whole IgG just above the hinge region to create two separate Fab fragments and one Fc fragment per antibody molecule. Because the papain protease is supplied in immobilized form as beaded agarose resin, the digestion reaction is easily stopped by removing the resin from the IgG solution; the result is digest products that are enzyme-free.

Related Products

  • Pierce™ Fab Micro Preparation Kit (Cat. No. 44685)
  • Pierce™ F(ab')2 Preparation Kit (Cat. No. 44988)
  • Pierce™ Mouse IgG1 Fab and F(ab')2 Preparation Kit (Cat. No. 44980)

Specifications

Content And Storage Sufficient For: 10 antibody samples, each containing 0.25 to 4 mg IgG
• Pierce Papain Agarose, 1.25 mL
• Cysteine-HCl, 1 g
• Fab Digestion Buffer, 120 mL
• NAb Protein A Plus Spin Column, 1 mL, 1 column
• PBS Packs (each makes 500 mL), 2 packs
• IgG Elution Buffer, 120 mL
• Zeba Desalt Spin Columns, 7K MWCO, 2mL, 10 columns
• Spin Columns, 0.8 mL, 10 columns
• Microcentrifuge Tubes, 2 mL, 30 tubes

Store at 4–8°C.
Product Line Pierce
Quantity 10 Antibody Samples, each containing 0.25 to 4 mg IgG kit
Can I reuse the immobilized digestion enzymes in the Thermo Scientific IgG Fragmentation kit?

No. For best results, use the immobilized enzyme once and discard after use.

How long does IgG digestion take using the Thermo Scientific IgG Fragmentation kit?

The digestion time is a function of the IgG concentration and source. For rabbit, mouse and human IgG, optimized digestion times are given in the instructions for each kit. Digestion times listed in the product instructions result in 80-90% digestion, although efficiency varies depending on IgG preparation and source.

In the Thermo Scientific IgG Fragmentation kit, when digesting an IgG that does not bind to Protein A, how do I purify the Fab and F(abÁ)2 fragments?

If the IgG is a species or subclass that does not bind to Protein A, researchers can immobilize an antibody specific for the desired fragment. For example, to purify digested rat IgG2a, use the Aminolink Plus Immobilization Kit (Cat. No. 44894) to immobilize goat anti-rat Fc antibody. Our Tech Tip #34 and other web resources list IgG binding properties of Protein A. (Because Protein G and Protein A/G bind Fab fragments, they are not effective for this application.)

Can I purchase the immobilized enzymes (proteases) from the Thermo Scientific IgG Fragmentation kit separately?

Yes. The three immobilized enzymes are available in 5 mL package sizes as shown below:

• Immobilized Ficin, Cat. No. 44881
• Immobilized Papain, Cat. No. 20341
• Immobilized Pepsin, Cat. No. 20343

How many Thermo Scientific IgG Fragmentation kits are available and which enzymes are included?

There are six Thermo Scientific IgG fragmentation kits:

• Fab Preparation Kit (Papain), Cat. No. 44985
• Fab Micro Preparation Kit (Papain), Cat. No. 44685
• F(abÁ)2 Preparation Kit (Pepsin), Cat. No. 44988
• F(abÁ)2 Micro Preparation Kit, (Pepsin), Cat. No. 44688
• Mouse IgG1 Fab and F(abÁ)2 Preparation Kit (Ficin), Cat. No. 44980
• Mouse IgG1 Fab and F(abÁ)2 Micro Preparation Kit (Ficin), Cat. No. 44680

What species of IgG can be fragmented with the Thermo Scientific IgG Fragmentation kits?

The protocols are optimized for rabbit, human and mouse IgG. Fragmentation of other IgG species may require optimization. To purify Fab or F(abÁ)2 fragments from the digest, the IgG species must be able to bind to Protein A. For mouse IgG1, use a ficin-based kit (Cat. Nos. 44980, 44680).


For Research Use Only. Not for use in diagnostic procedures.