missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ DAB Substrate Kit

Catalog No. PI34002
Change view
Click to view available options
Quantity:
275 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI34002 275 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI34002 Supplier Thermo Scientific™ Supplier No. LSG34002

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chromogenic horseradish peroxidase (HRP) substrate for Western blot and tissue staining methods.

Thermo Scientific™ Pierce DAB and the DAB Substrate Kit enable chromogenic detection of horseradish peroxidase (HRP) activity based the action of 3,3'-diaminobenzidine (DAB) in blotting and tissue staining methods.

DAB (3,3'-diaminobenzidine tetrahydrochloride; MW = 214.1) reacts with HRP in the presence of peroxide to yield an insoluble brown-colored product at locations where peroxidase-conjugated antibodies are bound to samples. The brown precipitate is insoluble in alcohol and other organic solvents, making it an excellent substrate for immunohistochemical staining that requires the use of traditional counterstains and mounting media. The Pierce DAB Substrate Kit includes a 10X solution of DAB and Stable Peroxide Substrate Buffer. Crystalline DAB (powder) is also available.

Highlights:

  • HRP substrate – for detection of horseradish peroxidase activity on solid media, including nitrocellulose and PVDF membranes and fixed tissue samples

  • Chromogenic – no special equipment needed for visualization; the DAB reaction produces brown bands or spots at sites of reaction with HRP-conjugated antibodies or probes

  • Package options – choose powder for custom formulation with buffer and peroxide or a complete kit of preformulated solutions for immediate use

Specifications

Description Pierce DAB Substrate Kit
Product Type Substrate
Detection Method Colorimetric
Form Liquid
Membrane Compatibility Nitrocellulose, PVDF
Product Line Pierce
Quantity 275 mL
Conjugate HRP
Substrate Horseradish Peroxidase
Reactivity Horseradish Peroxidase
Sufficient For 25 Uses at 10 mL Each
Content And Storage Sufficient For: 25 uses at 10 mL each
• DAB (10X), 25 mL
• Stable Peroxide Buffer, 250 mL

Store kit at 4°C.
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.