missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Cypridina-Firefly Luciferase Dual Assay Kit

Catalog No. p-4532161
Change view
Click to view available options
Quantity:
100 Reaction Kit
1000 Reaction Kit
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI16183 100 Reaction Kit
PI16184 1000 Reaction Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PI16183 Supplier Thermo Scientific™ Supplier No. 16183
Only null left

Simultaneously detect intracellular Cypridina and Red Firefly luciferase activity in mammalian whole cell lysates.

The Thermo Scientific™ Pierce Cypridina-Firefly Luciferase Dual Assay Kit provides the necessary reagents to simultaneously detect intracellular Cypridina and Red Firefly luciferase activity in mammalian whole cell lysates.

The Pierce Cypridina-Firefly Luciferase Dual Assay Kit is a highly sensitive assay that allows for the simultaneous detection of Cypridina and Red Firefly luciferase activity. Cypridina luciferase acts as an experimental reporter with constitutively active red firefly as a normalization control. This reporter-and-control combination enables simultaneous monitoring of experimental reporter and control luciferase activities in a single-read assay without the need for two-step addition of substrate reagents or quenching.

The assay working solution contains substrates for both luciferases, and the reactions occur simultaneously with flash-type kinetics. The resulting luminescent signals are spectrally resolvable using filters. In a single sample, researchers can assay transcriptional activity of regulatory elements, signal transduction pathways, and effects of activators or inhibitors.

Highlights

  • Simultaneous—concurrent, filter-based, wavelength-separated detection of two luciferase activities
  • Sensitive—measure bright blue Cypridina and red firefly luciferase activities in the same sample
  • Fast—no quenching step required, unlike in traditional sequential dual assays
  • Multiplex—capable of quantitating two cellular activities in the same sample(s)

Requires

Cypridina and Red Firefly luciferase reporters; filter sets in the ranges of 425 to 525 nm for Cypridina Luc and 615 to 675nm for Red Firefly Luc; luminometer or other instrument capable of monitoring luminescence, such as the Varioskan ALF or the Varioskan LUX microplate readers; injector for assessing more than 24 wells at a time

Recommended applications

Study two regulatory elements at the same time; monitor two signaling pathways simultaneously; enable studying more than one target per screen (e.g., off-target effects)

Specifications

Type Assay Kit
Target Luciferase, Cypridina Luciferase
Sufficient For 100 Dual Luciferase Assays
Quantity 100 Reaction Kit
Content And Storage Sufficient For: 100 dual-luciferase assays in microplate wells
• Cypridina-Firefly Dual Assay Buffer, 5 mL (store at 4°C)
• 100X Vargulin, 50 μL (store at -20°C)
• D-Luciferin, 3 mg (store at 4°C)
• Luciferase Cell Lysis Buffer (2X), 6 mL (store at room temperature)

Upon receipt, store kit at -20°C, or store individual components as indicated above.
Assay Reporter Enzyme, Luciferase Reporter Assay
Compatible Cells Mammalian Cells
Detection Method Bioluminescence
For Use With (Equipment) Luminometer (Microplate)
Format 384-well plate, 96-well plate
Label Type Enzyme Labeled
Substrate Properties Chemical Substrate
Technique Enhanced Chemiluminescence
Substrate Type Luciferase Substrate
Product Line Pierce
Substrate Vargulin (Cypridina Luciferin), D-Luciferin
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.