missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Chicken IgY Purification Kit

Catalog No. p-4527463
Change view
Click to view available options
Quantity:
1 L
5 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI44918 1 L
PI44922 5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PI44918 Supplier Thermo Scientific™ Supplier No. 44918
Only null left

Purify chicken antibodies from hen egg yolks with a unique delipidation reagent and precipitation reagent to selectively isolate the IgY fraction.

Thermo Scientific™ Pierce Chicken IgY Purification Kit purifies chicken antibodies from hen egg yolks using a unique delipidation reagent to separate protein from lipid and a precipitation reagent to selectively isolate the IgY fraction.

The IgY Purification Kit is simple and easy to use. After isolating a yolk using the supplied egg separator, lipids are effectively removed by incubation and centrifugation in Pierce Delipidation Reagent. This first reagent is also effective as a storage solution, enabling multiple yolks to be collected and stored for simultaneous extraction at a later date. Upon addition of the specialized Pierce IgY Precipitation Reagent and subsequent centrifugation, a nearly pure pellet of intact IgY results; it is easily dissolved in phosphate buffered saline (PBS) or other storage buffer. The purified IgY is ready for use in immunodetection methods or antigen-specific purification without interference from lipids.

Highlights:

  • Cost-effective – purifies twice as much IgY per egg than other popular kits, providing a lower cost-per-milligram of purified IgY

  • High purity – method yields chicken antibody that is 85 to 95% pure (based on SDS-PAGE analysis)

  • Easy to use – simple precipitation method purifies chicken IgY without affinity columns

  • Flexible protocol – eggs can be harvested and quickly stored in buffer for purification at a later date

  • Convenient – use eggs directly out of refrigerator; no need to equilibrate them to room temperature

Includes:

Egg separator, delipidation reagent and IgY precipitation reagent

Specifications

Product Type IgY Purification Kit
Product Line Pierce
Column Type Precipitation
Format Kit, Bottle
Purification Target IgY Antibody
Content And Storage Sufficient For: Purifying chicken IgY from 5 egg yolks
• Delipidation Reagent, 500 mL
• IgY Precipitation Reagent, 500 mL
• Egg Separator, 1 separator

Store at 4°C.
Quantity 1 L

For Research Use Only. Not for use in diagnostic procedures.