missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Bradford Plus Protein Assay Kits

Catalog No. PI23236
Change view
Click to view available options
Concentration:
BSA: 0.125, 0.25, 0.5, 0.75, 1, 1.5, and 2 mg/mL
BSA: 2 mg/mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Concentration
PI23236 BSA: 2 mg/mL
A55866 BSA: 0.125, 0.25, 0.5, 0.75, 1, 1.5, and 2 mg/mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PI23236 Supplier Thermo Scientific™ Supplier No. LSG23236

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

The Pierce Bradford Plus Protein Assay Kit is a ready-to-use, reducing agent-compatible, improved Bradford assay reagent used to quickly measure total protein concentration compared to a protein standard.

The Pierce Bradford Plus Protein Assay Kit is a ready-to-use, reducing agent-compatible, improved Bradford assay reagent used to quickly measure total protein concentration compared to a protein standard. It provides increased linearity and half the protein-to-protein variability of other commercial Bradford assay formulations.

Kits are available with or without Pierce Dilution-Free BSA Protein Standards, which are a set of seven pre-diluted BSA standards, packaged in a multichannel pipette-friendly tubestrip. The tubestrip includes a single empty tube that enables users to add their own sample buffer for the purpose of blank subtraction.

The Pierce Bradford Plus Protein Assay Kit is a ready-to-use, reducing agent-compatible, improved Bradford assay reagent used to quickly measure total protein concentration compared to a protein standard. It provides increased linearity and half the protein-to-protein variability of other commercial Bradford assay formulations.

Kit options include Pierce Dilution-Free BSA Protein Standards, which are a set of seven pre-diluted BSA standards, packaged in a multichannel pipette-friendly tubestrip. The tubestrip includes a single empty tube that enables users to add their own sample buffer for the purpose of blank subtraction.

Features of the Pierce Bradford Plus Protein Assay Kit include:

  • Colorimetric—measure with a standard spectrophotometer or plate reader at 595 nm
  • Easy to use—single reagent; no working reagent preparation required
  • Fast—almost immediate color development; add, mix, and read results
  • Wide assay range—detects protein concentration in the range 1 to 1500 μg/mL
  • Linear—improved linearity and response uniformity compared to traditional Bradford formulations
  • Flexible—microplate and cuvette protocols provided and adaptable to several target working ranges
  • Serial dilution–free—select kit options include Dilution-Free BSA Protein Standards that eliminate the need to perform serial dilutions when generating a standard curve

The assay is performed at room temperature. Simply add the sample to the tube or well containing reagent, and the resultant blue color is measured at 595 nm following a short room-temperature incubation. The protein assay is compatible with most salts, solvents, buffers, thiols, reducing substances, and metal-chelating agents encountered in protein samples. The assay can be performed in either test tube or microplate format.

How the Bradford Plus Assay detects protein

Use of Coomassie G-250 dye in a colorimetric reagent for the detection and quantitation of total protein was first described by Dr. Marion Bradford in 1976. In the acidic environment of the reagent, protein binds to the Coomassie dye. This results in a spectral shift from the reddish/brown form of the dye (absorbance maximum at 465 nm) to the blue form of the dye (absorbance maximum at 610 nm). The difference between the two forms of the dye is greatest at 595 nm, so that is the optimal wavelength to measure the blue color from the Coomassie dye-protein complex. If desired, the blue color can be measured at any wavelength between 575 nm and 615 nm. At the two extremes (575 nm and 615 nm) there is a loss of about 10% in the measured amount of color (absorbance) compared to that obtained at 595 nm.

Development of color in Coomassie dye-based (Bradford) protein assays has been associated with the presence of certain basic amino acids (primarily arginine, lysine, and histidine) in the protein. Van der Waals forces and hydrophobic interactions also participate in the binding of the dye by protein. The number of Coomassie dye ligands bound to each protein molecule is approximately proportional to the number of positive charges found on the protein. Free amino acids, peptides, and low molecular weight proteins do not produce color with Coomassie dye reagents. In general, the mass of a peptide or protein must be at least 3000 Daltons to be assayed with this reagent.

Specifications

Concentration BSA: 2 mg/mL
Content And Storage Sufficient For: 630 tube assays or 3160 microplate assays
• Pierce Bradford Plus Assay Reagent, 950 mL
• Albumin Standard ampules, 2 mg/mL, 10 x 1 mL

Store Bradford Plus Assay Reagent at 4°C. Store unopened Albumin Standard ampules at room temperature.
Assay Bradford Assay
Detection Method Colorimetric
Product Line Pierce
Product Type Protein Quantitation Assay
Specificity Not Target-Specific
Sufficient For 630 Tube Assays or 3160 Microplate Assays
For Use With (Application) Solution-based Detection, Absorbance
For Use With (Equipment) Spectrophotometer, Microplate Reader
Kit Contents 10 x 1 mL glass ampules
Quantity 950 mL
Show More Show Less
Which Thermo Scientific protein assay is the best or the most reliable?

The choice of protein assay is dependent on preferences related to assay speed, accuracy and sensitivity, as well as interfering substances in the sample to be assayed. BCA has less protein-to-protein variation, is compatible with most detergents, and has larger working range. Pierce Bradford Plus Protein Assay Kit (Cat. No. 23236) is compatible with reducing sugars, is more sensitive and is faster and easier to use. For a comparison of different protein assays and compatible reagents, see our Tech Tip: Protein Quantitation Assay Compatibility Table (https://assets.thermofisher.com/TFS-Assets/LSG/Application-Notes/TR0068-Protein-assay-compatibility.pdf).

What is the effect of temperature on the Pierce Bradford Plus Protein Assay Kit?

The absorbance readings at 595 nm obtained with the Pierce Bradford Plus Protein Assay Kit are dependent on the temperature of the reagent to some extent. As the reagent temperature increases to room temperature, the 595 nm readings will increase. Therefore, it is important that the Pierce Bradford Plus Protein Assay Reagent be at room temperature during the assay.

What is the detection range of Pierce Bradford Plus Protein Assay Kit?

Pierce Bradford Plus Protein Assay Kit has a detection range of 1µg/ml to 1,500µg/ml of protein. The protein of interest must have a molecular weight greater than 3,000 daltons to be detected by the assay. For peptides smaller than 3,000 daltons, try using Fluoraldehyde Reagent Solution (Cat. No. 26025), which will fluorescently detect amino groups on small peptide.

Other peptide quantitation assays we offer are Thermo Scientific Pierce Quantitative Colorimetric Peptide Assay (Cat. No. 23275) and Thermo Scientific Pierce Quantitative Fluorometric Peptide Assay (Cat. No. 23290).

What are the advantages of the Pierce Bradford Plus Protein Assay Kit over the Bradford Method?

Pierce Bradford Plus Protein Assay Kit has several advantages over the Bradford Method:

- The standard curve is more linear.
- No reagent preparation is required.
- No dilution or filtering is necessary.
- The reaction is very fast (results in as little as 30 seconds).

What agents are compatible with Pierce Bradford Plus Protein Assay Kit?

The assay is compatible with many agents that are incompatible in other assays. It has been shown to be compatible with amine-containing buffers, reducing agents, chaotropic agents, organic solvents, sugars, antimicrobial agents, DNA, protease inhibitors, and low concentrations of metal chelators and metals.

Is there any protein-to-protein variation when using the Pierce Bradford Plus Protein Assay Kit?

All proteins are unique and their varying structures can give variable responses. Each of the commonly used assay methods exhibits some degree of varying color response toward different proteins. These differences relate to dissimilarity among proteins due to amino acid sequence, pI, structure and the presence of certain side chains or prosthetic groups that can dramatically alter the protein's color response. Most protein assay methods utilize bovine serum albumin (BSA) or immunoglobulin (IgG) as the standard against which the concentration of protein in the sample is determined. Using either of these proteins as the standard works well in most assay methods. However, if great accuracy is required, the standard curve must be prepared from a pure sample of the target protein to be measured. If a pure sample of the target protein is not available, select the standard protein from those proteins that generate a color response that is close to the color response of the target protein.

How does the Pierce Bradford Plus Protein Assay Kit work?

The Pierce Bradford Plus Protein Assay Kit is based on an absorbance shift from 465 nm to 595 nm that occurs when the reagent binds to proteins in an acidic solution. The mechanism of the reaction is the anionic form of the dye complexes with proteins, resulting in a color change from brown to blue. The dye interacts chiefly with arginine residues and weakly with histidine, lysine, tyrosine, tryptophan and phenylalanine residues. The reaction reaches a stable endpoint so absorbance does not shift over time.

How can I remove interfering substances from my sample before using the Pierce Bradford Plus Protein Assay Kit?

You can remove interfering substances easily with the use of one of several Thermo Scientific products designed to purify your sample. These products include Slide-A-Lyzer Dialysis Cassettes, D-Salt Desalting Columns and Extracti-Gel D Detergent Removing Gel. Furthermore, TCA and cold acetone can be utilized to remove other interfering substances. (See the TechTips section of this web site for more information.)

Can detergents used to clean my glassware interfere with the Pierce Bradford Plus Protein Assay Kit?

Care must be exercised when cleaning glassware that will be used again for protein assays. Thorough cleaning often requires the use of a detergent, it must be completely removed in the final rinse. Also, the Coomassie dye will stain glass or quartz cuvettes. The cuvettes can be cleaned easily with our detergents followed by a thorough final rinsing with deionized water. If you prefer, disposable polystyrene cuvettes may be used to eliminate the cuvette-cleaning chore.

Can I read at wavelengths other than 595 nm with the Pierce Bradford Plus Protein Assay Kit?

If a photometer or plate reader is not available with a 595 nm filter, the blue color may be measured at any wavelength between 570 nm and 610 nm. The maximum sensitivity of the assay occurs when the absorbance of the Coomassie dye protein complex is measured at 595 nm. Taking the absorbance measurements at any wavelength other than 595 nm will result in a lower slope for the standard curve and may increase the minimum detection level for the protocol.

Can you provide the shelf life for Pierce Bradford Plus Protein Assay Kit?

Pierce Bradford Plus Protein Assay Kit is covered under our general 1-year warranty and is guaranteed to be fully functional for 12 months from the date of shipment, if stored as recommended. Please see section 8.1 of our Terms & Conditions of Sale (https://www.thermofisher.com/content/dam/LifeTech/Documents/PDFs/Terms-and-Conditions-of-Sale.pdf).

I assayed two protein samples, each containing a different mixture of proteins of same concentration and observed very different color responses in the assay. What is the cause?

Each of the commonly used total protein assay methods exhibits some degree of varying response toward different proteins. These differences relate to amino acid sequence, pI, structure and the presence of certain side chains or prosthetic groups that can dramatically alter the protein’s color response. Most protein assay methods use BSA or immunoglobulin (IgG) as the standard against which the concentration of protein in the sample is determined. However, if great accuracy is required, prepare the standard curve from a pure sample of the target protein.

My buffer or components of my buffer are not listed in the compatibility table for my protein assay. What should I do?

You can test the tolerance of the assay for your specific buffer formulation. For in-house generated compatibility information, substances were considered compatible at the indicated concentration in the Standard Test Tube Protocol (found in the manual for each protein assay) if the error in protein concentration estimation caused by the presence of the substance was less than or equal to 10%. The substances were tested using WR prepared immediately before each experiment. Blank-corrected 562nm absorbance measurements (for a 1000µg/mL BSA standard + substance) were compared to the net 562nm measurements of the same standard prepared in 0.9% saline.

All the components of my sample buffer are at or below the indicated compatible concentration for my protein assay, but I am still seeing too much/too little color development. What could be the problem?

It is possible to have a substance additive affect such that even though a single component is present at a concentration below its listed compatibility, a sample buffer containing a combination of substances could interfere with the assay. You should take steps to eliminate or minimize the effects of the interfering substance(s) by diluting or removing the substance.

Are there any reagents/chemicals I should avoid in my protein assay sample buffer?

Each protein assay method has its particular sensitivities. Generally, the following substances should be avoided for the given protein assay method:

  • BCA and Micro BCA Assays: reducing agents and chelators, and strong acids and bases
  • BCA Reducing Agent Compatible Assay: chelators 
  • 660 nm Assay: ionic detergents
  • Pierce Bradford Protein Assay Kit and Pierce Bradford Plus Protein Assay Kit: detergents
  • Modified Lowry Assay: detergents, reducing agents, and chelators 

A component of my lysis buffer is known to interfere with my protein assay of choice. How can I remove it?

Several strategies exist for overcoming or eliminating sample incompatibility with protein assays. The simplest method is to assay the sample after diluting it several-fold in a compatible buffer. If the starting concentration of protein is sufficient to remain within the protein assay working range upon its dilution, then this method will often reduce the amount of interfering substance in the sample to the point where it no longer interferes. Another method is to dialyze or desalt samples into a buffer that is compatible with the assay.

Precipitation can be used to eliminate interfering substances. After causing the protein to precipitate with acetone or trichloroacetic acid (TCA), the supernatant containing the interfering substance can be removed. Then the protein pellet is dissolved in the assay working reagent, and the protein assayed performed as usual. A general protocol for protein precipitation is provided in our Tech Tip. 

My protein assay is not developing color or is developing too much color. What can I do?

Refer to the information in the product-specific instruction booklet or our Tech Tip: Protein Quantitation Assay Compatibility Table (https://assets.thermofisher.com/TFS-Assets/LSG/Application-Notes/TR0068-Protein-assay-compatibility.pdf).

My spectrophotometer doesn’t have a filter set for the absorbance maximum. Can I use an alternate wavelength to read the protein assay?

Often, an alternative wavelength can be used, although the slope of the standard curve and the overall assay sensitivity will most likely be reduced. Our Tech Tip (https://tools.thermofisher.com/content/sfs/brochures/TR0025-Protein-assay-spectra.pdf) offers additional information on determining acceptable wavelengths for measuring protein assays.

Which protein assay is my lysis buffer compatible with?

Each method has its particular sensitivities. Apart from the information presented in the product-specific instruction booklet, we offer an expanded protein assay compatibility table in this Tech Tip.

What other factors affect the protein assay accuracy and precision?

Several factors affect protein assay accuracy and precision:
Replicates: The only way to evaluate the extent of random error is to include replicates of each standard and test sample. Because all test samples are evaluated by comparison to the standard curve, it is especially important to run the standards in at least triplicate. The standard deviation (SD) and coefficient of variation (CV) can then be calculated, providing a degree of confidence in your pipetting precision. If replicates are used, curve-fitting is done with the average values (minus obvious outliers).
Blank correction: It is common practice to subtract the absorbance of the zero assay standard(s) from the all other sample absorbance values. However, if replicate zero-assay standards will be used to calculate error statistics, then another independent value may be required for blank-correction. If the standards were prepared in a buffer to match that of the test samples, and this buffer contains components that may interfere with the assay chemistry, it is informative to blank the absorbances with a "water reference" (i.e., a zero-protein, water sample). Differences between the water reference and zero standard sample are then indicative of buffer effects.
Standard curve slope: The standard curve slope is directly related to assay accuracy and sensitivity. All else being equal, the steepest part of the curve is the most reliable. For most protein assays, the standard curve is steepest (i.e., has the greatest positive slope) in the bottom half of the assay range. In fact, the upper limit of an assay range is determined by the point at which the slope approaches zero; the line there is so flat that even a tiny difference in measured absorbance translates to a large difference in calculated concentration.
Measurement wavelength: The measurement wavelengths that are recommended for each protein assay method are optimal because they yield standard curves with maximal slope. This usually, but not always, corresponds to the absorbance maximum. (In certain circumstances, other considerations are also important in choosing the best possible measurement wavelength, such as avoiding interference from sample components that absorb at similar wavelengths). In fact, for most protein assays, depending on the precision required, acceptable results can be obtained using any measurement wavelengths within a certain range.

When does a dilution factor need to be applied in a protein assay?

One situation in which the dilution factor is important to consider is when the original sample has been pre-diluted relative to the standard sample. Suppose the original protein sample is actually known to be approximately 5 mg/mL. This is too concentrated to be assayed by the Pierce Bradford Plus Protein Assay Kit, for example, whose assay range in the standard microplate protocol is 100-1500 µg/mL. However, you could dilute it 5-fold in buffer (i.e., 1 part sample plus 4 parts buffer) and then use that diluted sample as the test sample in the protein assay. If the test sample produces the same absorbance as the 1000 µg/mL standard sample, then you can conclude that the test (5-fold diluted) sample is 1000 µg/mL, and therefore the original (undiluted) sample is 5 x 1000 µg/mL = 5000 µg/mL = 5 mg/mL.

Do I need to know the protein concentration in the assay reagent for my protein assay?

No. It is neither necessary nor helpful to know the protein concentration as it exists when the samples are diluted in assay reagent. The protein concentration when diluted by assay reagent is almost certainly not the value of interest; instead, one wants to know the protein concentration of the original test sample.

Do I need to know the amount of protein per well for my protein assay?

No. Contrary to what many people assume, it is neither necessary nor even helpful to know the actual amount (e.g., micrograms) of protein applied to each well or cuvette of the assay. The amount of protein per well is almost certainly not the value of interest; instead, one usually wants to know the protein concentration of the original test sample.

How can I utilize Excel software to plot and apply the standard curve for my protein assay?

Enter the concentration values for the standards in Column A and their corresponding absorbance data in Column B. Highlight both columns and from the Insert menu select Chart and XY (Scatter). Click on the resulting graph and select Add Trendline from the Chart menu. While viewing the graph next to the open Format Trendline window, choose Polynomial and set the Order to 2, 3 or 4 until the best-fit appears. Check the box near the bottom called Display Equation on Chart; then close the Format Trendline window. Use the resulting equation to determine protein concentration (y) of an unknown sample by inserting the sample’s absorbance value (x). 

How can I interpolate my protein assay data?

Most modern plate readers and spectrophotometers have associated software that automatically plots a linear or curvilinear regression line through the standard points, interpolates the test samples on that regression line, and reports the calculated value. However, there are different methods for making the calculations “by hand”. You can find a detailed explanation and example in our Tech Tip. 

How can I accurately analyze my protein assay data?

With most protein assays, sample protein concentrations are determined by comparing their assay responses to that of a dilution-series of standards whose concentrations are known. The responses of the standards are used to plot or calculate a standard curve. Absorbance values of unknown samples are then interpolated onto the plot or formula for the standard curve to determine their concentrations. The most accurate results are possible only when unknown and standard samples are treated identically. This includes assaying them at the same time and in the same buffer conditions, if possible. Because different pipetting steps are involved, replicates are necessary if you wish to calculate statistics (e.g., standard deviation, coefficient of variation) to account for random error. It is imperative to run a new standard curve for each set of samples to be tested

It was necessary to dilute my sample in order to run the protein assay (i.e,. due to an incompatible substance). How do I account for this when determining the concentration?

Simply multiply the calculated concentration of the diluted sample by the dilution factor. For example: A protein sample is known to be approximately 5 mg/mL. This is too concentrated to be assayed by the Pierce Bradford Plus Protein Assay Kit, whose assay range in the standard microplate protocol is 100-1500 µg/mL. However, you could dilute it 5-fold in buffer (i.e., 1 part sample plus 4 parts buffer) and then use that diluted sample as the test sample in the protein assay. If the test sample produces the same absorbance as the 1000 µg/mL standard sample, then you can conclude that the test (5-fold diluted) sample is 1000 µg/mL, and therefore the original (undiluted) sample is 5 × 1000 µg/mL = 5000 µg/mL = 5 mg/mL.

In my protein assay, what unit of measurement will my sample concentration be in after calculating the concentration?

The unit of measure used to express the standards is by definition the same unit of measure associated with the calculated value for the unknown sample (i.e., final results for unknown samples will be expressed in the same unit of measure as was used for the standards). For example, if the standard concentrations are expressed as micrograms per milliliter, then the concentrations for the unknown samples, which are determined by comparison to the standard curve, are also expressed as micrograms per milliliter.

Can I re-use my protein standard curve dilutions that I prepared?

Yes, we recommend storing at -20 degrees C and they will likely be good for 2-3 months, or about 2 freeze/thaws. Samples subjected to more than ˜2 freeze/thaw cycles may give variable results in most assays

What should I dilute my protein standard in for my protein assay?

Protein standards should preferably be diluted using the same diluent as the sample(s). Sample assay responses are directly comparable to each other if they are processed in exactly the same manner. Variance in protein quantity is the only possible cause for differences in final absorbance (color intensity) if samples are dissolved in the same buffer and the same stock solution of assay reagent is used for all samples.

However, if only a “rough” estimate of protein concentration is needed, a blank-only correction can be used. In this case, a blank is prepared in the diluent of the sample to correct for its raw absorbance. The concentration of the sample is then determined from a standard curve obtained from a series of dilutions of the protein of known concentration prepared in water or saline solution.

What proteins do you offer as standards for your protein assays?

We offer BSA and BGG as protein standards for protein assays.

What protein should I use to generate my standard curve?

Protein concentrations are generally determined and reported with reference to standards of a common protein, such as bovine serum albumin (BSA). If precise quantitation of an unknown protein is required, it is advisable to select a protein standard that is similar in quality to the unknown; for example, a bovine gamma globulin (BGG) standard may be used when assaying immunoglobulin samples.

Why is the choice of protein standard important in a protein assay?

Because proteins differ in their amino acid compositions, each one responds somewhat differently in each type of protein assay. Therefore, the best choice for a reference standard is a purified, known concentration of the most abundant protein in the samples. This is usually not possible to achieve, and it is seldom convenient or necessary. If a highly purified version of the protein of interest is not available or it is too expensive to use as the standard, the alternative is to choose a protein that will produce a very similar color response curve in the selected protein assay method and is readily available to any laboratory at any time. Generally, bovine serum albumin (BSA) works well as a protein standard because it is widely available in high purity and relatively inexpensive. Alternatively, bovine gamma globulin (BGG) is a good standard when determining the concentration of antibodies because BGG produces a color response curve that is very similar to that of immunoglobulin G (IgG).

What are the basic principles of standard curve assays?
  • Identically assayed samples are directly comparable: Sample assay responses are directly comparable to each other if they are processed in exactly the same manner. Variation in amount of protein is the only possible cause for differences in final absorbance (color intensity) if the samples are dissolved in the same buffer, the same lot and stock solution of assay reagent is used, all samples are mixed and incubated at the same time and temperature, and no pipetting errors were introduced. 
  • Units in equals the units out: The unit of measure used to express the standards is by definition the same unit of measure associated with the calculated value for the unknown sample (i.e., final results for unknown samples will be expressed in the same unit of measure as was used for the standards).

Which protein assays are dye-binding based chemistries?

Pierce Bradford Protein Assay Kit and Pierce Bradford Plus Protein Assay Kit are variations on the use of Coomassie G-250 dye as a colorimetric reagent for the detection and quantitation of total protein first reported by Bradford in 1976. The Thermo Scientific 660 nm Protein Assay is a dye-based reagent that offers the same convenience as Coomassie-based assays while overcoming several of their disadvantages. In particular, the 660 nm Assay is compatible with most detergents and produces a more linear response curve (the detailed assay chemistry is proprietary). Our fluorometric protein assays are also based on dye binding chemistries.

What is the chemistry behind protein assays?

Most colorimetric protein assay methods can be divided into two groups based on the type of chemistry involved: those involving protein-copper chelation with secondary detection of the reduced copper and those based on protein-dye binding with direct detection of the color change associated with the bound dye.

What is protein-to-protein variation?

Each protein in a sample responds uniquely in a given protein assay, and this protein-to-protein variation is observed as differences in the amount of color (absorbance) obtained when the same mass of various proteins is assayed concurrently by the same method. These differences in color response relate to differences in amino acid sequence, isoelectric point (pI), secondary structure, and the presence of certain side chains or prosthetic groups.

Depending on the sample type and purpose for performing an assay, protein-to-protein variation is an important consideration in selecting a protein assay method and in selecting an appropriate assay standard (e.g., BSA vs. BGG). Protein assay methods based on similar chemistry have similar protein-to-protein variation.

How should a sample be prepared before a protein assay?

Before the sample is analyzed, it must be solubilized in a buffered aqueous solution. Depending on the source material and the procedures involved before performing the protein assay, the sample will likely contain a variety of non-protein components. Awareness of these components is critical for choosing an appropriate assay method and evaluating the cause of anomalous results. Every type of protein assay is adversely affected by substances of one sort or another. Components of a protein solution are considered interfering substances in a protein assay if they artificially suppress the response, enhance the response, or cause elevated background by an arbitrarily chosen degree (e.g., 10% compared to control). Additional components can include reducing agents, chelators, crowding agents, and protease inhibitors.

What should I consider when choosing a protein assay?

There are several criteria that should be considered, including compatibility with the sample type and components, assay range and required sample volume, protein-to-protein uniformity, speed and convenience for the number of samples to be tested, and the availability of spectrophotometer or plate reader necessary to measure the color produced (absorbance) by the assay.

Why does the protein assay method matter?

Unfortunately, no protein assay method exists that is either perfectly specific to proteins (i.e., not affected by any nonprotein components) or uniformly sensitive to all protein types (i.e., not affected by differences in protein composition). Therefore, successful use of protein assays involves selecting the method that is most compatible with the samples to be analyzed, choosing an appropriate assay standard, and understanding and controlling the particular assumptions and limitations that remain. The objective is to select a method that requires the least manipulation or pre-treatment of the samples to accommodate substances that interfere with the assay. Each method has its particular advantages and disadvantages. Because no one reagent can be considered the ideal or best protein assay method for all circumstances, most researchers have more than one type of protein assay available in their laboratories.

What protein assay is best?

Unfortunately, no protein assay method exists that isn’t affected by any non-protein component or uniformly sensitive to all protein types. One must select an appropriate assay method based on compatibility with the sample type or one that requires the least manipulation of the sample to accommodate the assay. Most researchers will have more than one type of assay available in their laboratories.

What protein assays do you offer for total protein quantitation?

We offer several types of protein assays including the: BCA Assay, BCA-RAC (Reducing Agent Compatible) Assay, Micro BCA Assay, 660 nm Protein Assay, Pierce Bradford Plus Protein Assay Kit, Pierce Bradford Protein Assay Kit, Modified Lowry Assay, colorimetric and fluorometric Peptide Assays, CBQCA kit, EZQ kit, Quant-iT kits, NanoOrange, and the Qubit kits.

What is the difference between the Pierce Bradford Protein Assay Kit and the Pierce Bradford Plus Protein Assay Kit?

Pierce Bradford Plus Protein Assay Kit has all the advantages of the regular Pierce Bradford Protein Assay plus the added benefit of having a more linear standard curve. The added linearity of the standard curve translates to higher accuracy in the protein assay.

Which protein assay is my lysis compatible with?

Please check the Protein Assay Compatibility Table in this Tech Tip (https://assets.thermofisher.com/TFS-Assets/LSG/Application-Notes/TR0068-Protein-assay-compatibility.pdf) to determine which protein assay would be most effective with your sample type.


For Research Use Only.