missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ BCA Protein Assay Kits

Catalog No. p-200000333
Change view
Click to view available options
Quantity:
1 Kit (1 L)
1 Kit (100 mL)
1 Kit (500 mL)
Concentration:
BSA: 0.125, 0.25, 0.5, 0.75, 1, 1.5, and 2 mg/mL
BSA: 2 mg/mL
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Concentration
FERA65453 1 Kit (100 mL) BSA: 2 mg/mL
PI23225 1 Kit (1 L) BSA: 2 mg/mL
PI23227 1 Kit (500 mL) BSA: 2 mg/mL
A55864 1 Kit (1 L) BSA: 0.125, 0.25, 0.5, 0.75, 1, 1.5, and 2 mg/mL
A55865 1 Kit (500 mL) BSA: 0.125, 0.25, 0.5, 0.75, 1, 1.5, and 2 mg/mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. FERA65453 Supplier Thermo Scientific™ Supplier No. A65453

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

The Pierce BCA Protein Assay Kit is a high-precision, detergent-compatible protein assay for determination of protein concentration. Pierce BCA reagents provide accurate determination of protein concentration with most sample types encountered in protein research.

The Pierce BCA Protein Assay Kit is a high-precision, detergent-compatible protein assay for determination of protein concentration. Pierce BCA reagents provide accurate determination of protein concentration with most sample types encountered in protein research. The Pierce BCA assay can be used to assess yields in whole cell lysates, affinity-column fractions, purified proteins samples, as well as to monitor protein contamination in industrial applications. Compared to most dye-binding methods, the BCA assay is affected much less by protein compositional differences, providing greater concentration accuracy.

Kits are available with or without Pierce Dilution-Free BSA Protein Standards, which are a set of seven pre-diluted BSA standards, packaged in a multichannel pipette-friendly tubestrip. The tubestrip includes a single empty tube that enables users to add their own sample buffer for the purpose of blank subtraction.

The Pierce BCA Protein Assay Kit is a high-precision, detergent-compatible protein assay for determination of protein concentration. Pierce BCA reagents provide accurate determination of protein concentration with most sample types encountered in protein research. The Pierce BCA assay can be used to assess yields in whole cell lysates, affinity-column fractions, purified proteins samples, as well as to monitor protein contamination in industrial applications. Compared to most dye-binding methods, the BCA assay is affected much less by protein compositional differences, providing greater concentration accuracy.

Kit options are available that include Pierce Dilution-Free BSA Protein Standards, which are a set of seven pre-diluted BSA standards, packaged in a multichannel pipette-friendly tubestrip.  The tubestrip includes a single empty tube that enables users to add their own sample buffer for the purpose of blank subtraction.

Features of the BCA Protein Assay Kit include:

  • Colorimetric—estimate visually or measure with a standard spectrophotometer or plate reader at 562 nm
  • Accurate—exhibits half the protein-to-protein variation observed with dye-binding methods (Bradford)
  • Compatible—unaffected by typical concentrations of most ionic and non-ionic detergents
  • Short assay time—30-min incubation; much easier and four times faster than classical Lowry methods
  • Wide assay range—linear working range for BSA of 20 to 2000 μg/mL
  • Sensitive—detect down to 5 μg/mL with enhanced protocol
  • No more serial dilutions—kit options include Dilution-Free BSA Protein Standards which eliminate the need to perform serial dilutions when generating a standard curve

How the assay works

The BCA Protein Assay combines the well-known reduction of Cu2+ to Cu1+ by protein in an alkaline medium with the highly sensitive and selective colorimetric detection of the cuprous cation (Cu1+) by bicinchoninic acid (BCA). The first step is the chelation of copper with protein in an alkaline environment to form a light blue complex. In this reaction, known as the biuret reaction, peptides containing three or more amino acid residues form a colored chelate complex with cupric ions in an alkaline environment containing sodium potassium tartrate.

In the second step of the color development reaction, BCA reacts with the reduced (cuprous) cation that was formed in step one. The intense purple-colored reaction product results from the chelation of two molecules of BCA with one cuprous ion. The BCA/copper complex is water soluble and exhibits a strong linear absorbance at 562 nm with increasing protein concentrations. The complex is approximately 100 times more sensitive (lower limit of detection) than the pale blue color of the first reaction.

The reaction is strongly influenced by four amino acid residues (cysteine, cystine, tyrosine, and tryptophan) in the amino acid sequence of the protein. However, unlike Coomassie dye-binding methods (i.e., Bradford), the universal peptide backbone also contributes to color formation, helping to minimize variability caused by protein compositional differences.

Specifications

Concentration BSA: 2 mg/mL
Content And Storage Sufficient For: 50 tube assays or 500 microplate assays
• BCA Reagent A, 100 mL
• BCA Reagent B, 2.5 mL
• Albumin Standard, 2 mg/mL, 2 mL

Store at room temperature. Store BSA Protein Standards at 4°C after opening.
Assay BCA Assay
Detection Method Colorimetric
Product Line Pierce
Product Type Protein Quantitation Assay
Specificity Not Target-Specific
Sufficient For 50 Tube Assays or 500 Microplate Assays
For Use With (Application) Solution-based Detection, Absorbance
For Use With (Equipment) Spectrophotometer, Microplate Reader
Kit Contents 2 mL vial
Quantity 1 Kit (100 mL)
Show More Show Less
Which Thermo Scientific protein assay is the best or the most reliable?

The choice of protein assay is dependent on preferences related to assay speed, accuracy and sensitivity, as well as interfering substances in the sample to be assayed. BCA has less protein-to-protein variation, is compatible with most detergents, and has larger working range. Pierce Bradford Plus Protein Assay Kit (Cat. No. 23236) is compatible with reducing sugars, is more sensitive and is faster and easier to use. For a comparison of different protein assays and compatible reagents, see our Tech Tip: Protein Quantitation Assay Compatibility Table (https://assets.thermofisher.com/TFS-Assets/LSG/Application-Notes/TR0068-Protein-assay-compatibility.pdf).

What substances interfere with the BCA Protein Assay?

Reducing agents, copper chelators and solutions with very high buffering capacities will interfere with the BCA assay. Reducing agents reduce the copper and will produce a high background. Copper chelators bind the copper and prevent it from being detected by the BCA reagent. High-capacity buffers prevent the BCA from reaching its optimal alkaline pH. For a complete list of compatible substances, please refer to the BCA Protein Assay Kit product instructions.

What is the sample to working reagent ratio when using the BCA Protein Assay Kit?

For the Microplate Protocol the sample to working reagent (WR) ratio is 1:8 (25 µl sample plus 200 µl WR). If the sample amount is limited, a 1:20 ratio may be used (10 µl sample); in this case, however, the detection range of the assay will be limited to 125-2,000 µg/ml. For the Test Tube Procedure (Standard or Enhanced Protocols) the ratio is 1:20 (0.1 ml sample plus 2.0 ml WR). In the Micro BCA Assay Kit (for both Microplate and Test Tube Procedures), the ratio is 1:1 (1 part sample plus 1 part WR).

What is the detection range for the BCA Protein Assay?

For the standard protocol, the detection range is 20-2,000 µg/ml. For the enhanced test tube protocol, the detection range is 5-250 µg/ml. The Micro BCA Protein Assay Reagent will detect 0.5-20 µg/ml of protein in the test tube assay and 1-20 µg/ml for the microplate assay.

How can interfering substances be eliminated from the protein sample when using the BCA Protein Assay Kit?

There are several ways to adjust a sample to be compatible with the BCA Protein Assay: • Remove the interfering substance by dialysis or gel filtration. • If the starting protein concentration of the sample is high, dilute the sample to the point that the substance no longer interferes. • Eliminate interference by copper-chelating agents by increasing the amount of copper in the working reagent (use 4 ml or 6 ml of Reagent B/100 ml of Reagent A instead of the 2 ml of Reagent B/100 ml of Reagent A that is called for in the instructions). • Precipitate sample proteins with cold acetone or trichloroacetic acid (TCA).

Is a specific microplate recommended for the BCA Protein Assay?

Although the brand of plate should not affect the BCA Protein Assay, assay results may be affected by well configuration. The microplate protocol is based on the use of clear, flat-bottomed plates. The recommended plates for use with the BCA Protein Assay are Pierce 96-Well Polystyrene Plates, Corner Notch (Cat. No. 15041). If these plates are not available, other clear flat-bottomed plates are acceptable as the plate surface chemistry will not alter BCA Protein Assay results. Using round-bottomed plates may affect the A(562) readings because the plate reader may not be able to align the plate within the optical path with reproducibility, which will cause variation in the absorbance from well to well. To achieve optimum results, Thermo Fisher Scientific recommends the use of new plates and glassware that have been thoroughly cleaned and rinsed before use.

Do I need to make a standard curve each time I perform the BCA Protein assay?

For optimal accuracy, make a standard curve each time a protein assay is performed. Measure standards in duplicate for the test tube assay and in triplicate for the plate assay.

Can the BCA Protein Assay Kit quantitate peptides?

Peptides smaller than 2,000 Da cannot be quantitated with the BCA assay because color formation is dependent on the protein's macromolecular structure, the number of peptide bonds, and the presence of cysteine, cystine, tryptophan and tyrosine. For peptides smaller than 2,000 Da, try using Fluoraldehyde o-Phthalaldehyde Reagent Solution (Cat. No. 26025). This reagent will detect amino groups on small peptides by fluorescence.

Other peptide quantitation assays we offer are Thermo Scientific Pierce Quantitative Colorimetric Peptide Assay (Cat. No. 23275) and Thermo Scientific Pierce Quantitative Fluorometric Peptide Assay (Cat. No. 23290).

At what wavelength is the BCA Protein Assay measured?

The maximum absorbance of the BCA-Cu+1 complex occurs at 562 nm. If a spectrophotometer or plate reader with a 562 nm filter is not available, the purple color may be measured at any wavelength between 540 nm and 590 nm. Measuring the absorbance at any wavelength other than 562 nm will result in a lower slope for the standard curve and may increase the minimum detection level for the protocol.

How does the Pierce BCA Protein Assay Kit work?

The Pierce BCA Protein Assay combines the well-known reduction of Cu2+ to Cu1+ by protein in an alkaline medium with the highly sensitive and selective colorimetric detection of the cuprous cation (Cu1+) by bicinchoninic acid (BCA). The first step is the chelation of copper with protein in an alkaline environment to form a light blue complex. In this reaction, known as the biuret reaction, peptides containing three or more amino acid residues form a colored chelate complex with cupric ions in an alkaline environment containing sodium potassium tartrate.

In the second step of the color development reaction, BCA reacts with the reduced (cuprous) cation that was formed in step one. The intense purple-colored reaction product results from the chelation of two molecules of BCA with one cuprous ion. The BCA/copper complex is water soluble and exhibits a strong linear absorbance at 562 nm with increasing protein concentrations. The complex is approximately 100 times more sensitive (lower limit of detection) than the pale blue color of the first reaction.

The reaction is strongly influenced by four amino acid residues (cysteine, cystine, tyrosine, and tryptophan) in the amino acid sequence of the protein. However, unlike Coomassie dye-binding methods (i.e., Bradford), the universal peptide backbone also contributes to color formation, helping to minimize variability caused by protein compositional differences.

What does the maximum concentrations in the Protein quantitation assay compatibility table for the Pierce BCA Protein Assay Kit (Cat. Nos. 23225, 33227) refer to?

The maximum concentrations in the Protein quantitation assay compatibility table for the Pierce BCA Protein Assay Kit (Cat. Nos. 23225, 23227) refers to the actual concentration in the protein sample before any working reagent is added.

What BCA protein assay kits do you recommend for NanoDrop 2000/2000c Spectrophotometers (Cat. Nos. ND-2000, ND-2000C)?

NanoDrop 2000/2000c Spectrophotometers (Cat. Nos. ND-2000, ND-2000C) are compatible with most commercial BCA protein assay kits. We offer a number of BCA protein assay kits, including, but not limited to, the BCA Protein Assay Kit (Cat. No. J63283.QA), Micro BCA Protein Assay Kit (Cat. No. 23235, and Pierce BCA Protein Assay Kit (Cat. No 23225, 23227).

To identify, compare, and choose the best protein assay for your sample type and application, you can use our interactive Protein Assay Selection Guide.

Is the Pierce BCA Protein Assay Kit (Cat. No. 23225, 23227) compatible with OptiPrep (Iodixanol)?

We have not tested the Pierce BCA Protein Assay Kit compatibility with OptiPrep (Iodixanol), therefore we cannot recommend using them together.

Can you provide the shelf-life for the Pierce BCA Protein Assay Kit?

The Pierce BCA Protein Assay Kit is covered under our general 1-year warranty and is guaranteed to be fully functional for 12 months from the date of shipment, if stored as recommended (room temperature). Please see section 8.1 of our Terms & Conditions of Sale (https://www.thermofisher.com/content/dam/LifeTech/Documents/PDFs/Terms-and-Conditions-of-Sale.pdf) for more details.

I am using the Pierce BCA Protein Assay Kit (Cat. No. 23225) and have made all the standards in my assay. Can I store the standards at room temperature and if so, for how long?

The standards can be stored overnight at room temperature or for up to a week at 4 degrees C if they are in a tightly closed container.

Does Y-PER Yeast Protein Extraction Reagent (Cat. No. 78990) yield samples that are compatible with the Pierce BCA Protein Assay Kit (Cat. No. 23225)?

Yes, Y-PER Yeast Protein Extraction Reagent is compatible with our Pierce BCA Protein Assay Kit. Please see our Protein assay compatibility table in this Tech Tip (https://assets.thermofisher.com/TFS-Assets/LSG/Application-Notes/TR0068-Protein-assay-compatibility.pdf).

When I use Pierce BCA Protein Assay Kit, what if my spectrophotometer does not have a 562 nm filter?

You can measure the color at wavelengths between 540nm and 590 nm, although the slope for the standard curve and overall assay sensitivity is reduced.

When I use Pierce BCA Protein Assay Kit, why are all of the tubes, including the blank, dark purple?

Dark purple coloring can indicate that the buffer contains a reducing agent, such as a thiol or biogenic amines. We recommend either dialyzing or diluting the sample. Alternatively, you could remove the interfering substance using Compat-Able Protein Assay Preparation Reagent Kit (Cat. No. 23215).

When I use Pierce BCA Protein Assay Kit, why is the color of the samples darker than expected?

Darker samples could indicate that the protein concentration is too high, which can be corrected by diluting the sample. The sample could also contain lipids or lipoproteins. We recommend adding 2% SDS to eliminate interference from lipids. Alternatively, you could remove interfering substances using Compat-Able Protein Assay Preparation Reagent Kit (Cat. No. 23215).

When using a Pierce BCA Protein Assay Kit, why is my blank correct, but the standards and samples show less color than expected?

The sample could contain a strong acid or alkaline buffer, which could alter the working reagent pH. If this occurs, we recommend dialyzing, desalting, or dilute the sample.
Alternatively, the color may have been measured at the wrong wavelength. In this case, we recommend measuring the absorbance 562 nm.

Why is there no color in my tubes from the Pierce BCA Protein Assay Kit?

The sample could contain a copper chelating agent.
We recommend trying one of the following possible solutions:
- Dialyze, desalt, or dilute the sample
- Increase the copper concentration in the working reagent (use 50: Reagent A:B)
- Remove interfering substances from the sample using the Compat-Able Protein Assay Preparation Reagent Kit (Cat. No. 23215)

How do I determine the coupling efficiency of my protein to a solid support?

Immobilized protein samples can be quantified using Pierce BCA Protein Assay or Coomassie Plus Protein Assay. Please refer to this Tech Tip on how to "Quantitate Immobilized Protein"(http://tools.thermofisher.com/content/sfs/brochures/TR0009-Quant-immobilized-protein.pdf) . Alternatively, this amount can be estimated by measuring the protein concentration in solution before and after coupling. The difference in the two measurements is the amount that is bound to the support

Which protein assay is my lysis compatible with?

Please check the Protein Assay Compatibility Table in this Tech Tip (https://assets.thermofisher.com/TFS-Assets/LSG/Application-Notes/TR0068-Protein-assay-compatibility.pdf) to determine which protein assay would be most effective with your sample type.


For Research Use Only.