Learn More
Thermo Scientific™ Pierce™ Avidin
Description
Thermo Scientific™ Pierce Avidin and Conjugates include purified avidin protein from hen egg whites, plus avidin-HRP, avidin-AP and other conjugated forms for use in biotin-based detection methods.
Highlights:
Native avidin – glycoprotein from chicken egg whites, 67kDa, pI equals 10
Soluble – glycosylation and isoelectric point renders avidin more soluble than streptavidin
Affordable – significantly less expensive than recombinant streptavidin
ABC staining – avidin is usually preferred over streptavidin for avidin-biotin complex (ABC) staining methods in immunohistochemistry (IHC)
Pure protein and conjugates – choose pure, unconjugated avidin for use in ABC staining or peroxidase or alkaline phosphatase forms for membrane and plate-based assays
Specifications
Specifications
| Conjugate | Unconjugated |
| Quantity | 10 mg |
| Content And Storage | Store at -20°C. Shipped at ambient temperature. |
| Form | Solid |
| Product Line | Pierce |
| Product Type | Avidin (Unconjugated) |
| Target Molecule | Biotin |
| Label or Dye | Unlabeled |
For Research Use Only. Not for use in diagnostic procedures.