missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Avidin

Catalog No. PI21128
Change view
Click to view available options
Quantity:
10 mg
20 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI21128 20 mg
PI21121 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PI21128 Supplier Thermo Scientific™ Supplier No. 21128

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Perform Western blot, ELISA or other biotin-based detection methods using these purified and HRP-, AP- or FITC-labeled avidin molecules.

Thermo Scientific™ Pierce Avidin and Conjugates include purified avidin protein from hen egg whites, plus avidin-HRP, avidin-AP and other conjugated forms for use in biotin-based detection methods.

Avidin from chicken egg whites is a glycoprotein that effectively binds biotin, a tag that is frequently used to tag antibodies and other probes for immunodetection methods. Pure avidin is a key component in certain immunohistochemistry (IHC) methods. Alkaline phosphatase (AP) and horseradish peroxidase (HRP) conjugates of avidin are useful for Western blotting or ELISA detection with appropriate enzyme substrates.

Highlights:

  • Native avidin – glycoprotein from chicken egg whites, 67kDa, pI equals 10

  • Soluble – glycosylation and isoelectric point renders avidin more soluble than streptavidin

  • Affordable – significantly less expensive than recombinant streptavidin

  • ABC staining – avidin is usually preferred over streptavidin for avidin-biotin complex (ABC) staining methods in immunohistochemistry (IHC)

  • Pure protein and conjugates – choose pure, unconjugated avidin for use in ABC staining or peroxidase or alkaline phosphatase forms for membrane and plate-based assays

Specifications

Conjugate Unconjugated
Quantity 20 mg
Content And Storage Store at -20°C. Shipped at ambient temperature.
Form Solid
Product Line Pierce
Product Type Avidin (Unconjugated)
Target Molecule Biotin
Label or Dye Unlabeled

For Research Use Only. Not for use in diagnostic procedures.