missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Amino Acid Standard H

Catalog No. PI20088
Change view
Click to view available options
Quantity:
10 x 1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI20088 10 x 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI20088 Supplier Thermo Scientific™ Supplier No. 20088

missing translation for 'validMainframeMsg'

Calibrate HPLC analysis of protein hydrolysates using this calibration standard mixture of 18 amino acids, each at 2.5 micromoles/mL in 0.1N HCl.

Thermo Scientific™ Pierce Amino Acid Standard H is a quantitative mixture of 18 amino acids, supplied at 2.5μmol/mL in 0.1N HCl, for use as a high-purity calibration standard for HPLC analysis of protein hydrolysates.

Highlights:

  • Uses L-form configuration to permit standardization of microbial and other assays
  • Molar concentration verified by conventional amino acid analysis methods
  • With the exception of cystine, each amino acid is supplied at a concentration of 2.5μmoles/mL in 0.1N HCl
  • L-alanine, Ammonia [(NH4)2SO4], L-Arginine, L-Aspartic Acid, L-Cystine, L-Glutamic Acid, Glycine, L-Histidine, L-Isoleucine, L-Leucine, L-Lysine·HCl, L-Methionine, L-Phenylalanine, L-Proline, L-Serine, L-Threonine, L-Tyrosine, L-Valine

Includes:

  • L-alanine, Ammonia [(NH4)2SO4], L-Arginine, L-Aspartic Acid, L-Cystine, L-Glutamic Acid, Glycine, L-Histidine, L-Isoleucine, L-Leucine, L-Lysine·HCl, L-Methionine, L-Phenylalanine, L-Proline, L-Serine, L-Threonine, L-Tyrosine, L-Valine

Specifications

Product Type Amino Acid Standard H
Content And Storage Store between -20°C and 8°C.
Quantity 10 x 1 mL
Form Liquid

For Research Use Only. Not for use in diagnostic procedures.