missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Alkaline Phosphatase

Purified calf-intestinal alkaline phosphatase (AP, alk-phos) enzyme for use in activity assays and conjugation to antibodies for ELISA and IHC.

Supplier:  Thermo Scientific™ 31392

 View more versions of this product

Catalog No. PI31392


Only null left
Explore available promotions

Description

Description

Thermo Scientific™ Pierce Alkaline Phosphatase is purified calf-intestinal alkaline phosphatase (AP or alk-phos) enzyme for use in activity assays and conjugation to antibodies for ELISA and immunohistochemistry applications.

Highlights:

  • Purified form - ready to conjugate without prior dialysis
  • Activity is not affected by exposure to antibacterial agents, such as sodium azide or thimerosal
  • Specific activity > 2,000 units/mg
  • One unit is defined as the amount that will hydrolyze 1.0 mol of p-nitrophenyl phosphate per minute at 37° C in 1.0M diethanolamine, 0.5mM MgCl2, pH 7.8
Specifications

Specifications

Store at 2–8°C.
ELISA, Immunohistochemistry
AP (Alkaline Phosphatase)
Liquid
Pierce
5 x 20 mg
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

For Research Use Only. Not for use in diagnostic procedures.