missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Alkaline Phosphatase

Catalog No. PI31391
Change view
Click to view available options
Quantity:
20 mg
5 x 20 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI31391 20 mg
PI31392 5 x 20 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PI31391 Supplier Thermo Scientific™ Supplier No. 31391

Purified calf-intestinal alkaline phosphatase (AP, alk-phos) enzyme for use in activity assays and conjugation to antibodies for ELISA and IHC.

Thermo Scientific™ Pierce Alkaline Phosphatase is purified calf-intestinal alkaline phosphatase (AP or alk-phos) enzyme for use in activity assays and conjugation to antibodies for ELISA and immunohistochemistry applications.

Highlights:

  • Purified form - ready to conjugate without prior dialysis
  • Activity is not affected by exposure to antibacterial agents, such as sodium azide or thimerosal
  • Specific activity > 2,000 units/mg
  • One unit is defined as the amount that will hydrolyze 1.0 mol of p-nitrophenyl phosphate per minute at 37° C in 1.0M diethanolamine, 0.5mM MgCl2, pH 7.8

Specifications

Content And Storage Store at 2–8°C.
Form Solid
For Use With (Application) ELISA, Immunohistochemistry
Product Line Pierce
Label or Dye AP (Alkaline Phosphatase)
Quantity 20 mg

For Research Use Only. Not for use in diagnostic procedures.