missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Active Rac1 Pull-Down and Detection Kit

Catalog No. PI16118
Change view
Click to view available options
Quantity:
30 reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI16118 30 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI16118 Supplier Thermo Scientific™ Supplier No. 16118

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Purify GTPases and assay activation levels using specific GST-tagged protein-binding domains (PBD), GSH-agarose beads and anti-GTPase antibodies.

Measure activation of small GTPases by isolating them via their specific downstream effectors with Thermo Scientific™ Pierce Active GTPase Pull-Down and Detection Kits.

The Active GTPase Pull-Down and Detection Kits measure activation of small GTPases by isolating them via their specific downstream effectors. Kits specific for Arf1, Arf6, Cdc42, Rac1, RalA, Rap1, Ras, and Rho GTPases are offered. The respective binding domain of the downstream effector for each small GTPase is expressed as a GST-fusion protein. When immobilized on a resin, this fusion protein is used to pull down the active (i.e., GTP-bound) GTPase. The pull-down affinity purification step is easily and conveniently performed using microcentrifuge spin columns. After a simple wash step, the bound GTPase is recovered by eluting the GST-fusion protein from the glutathione resin using SDS-PAGE sample loading buffer. The purified GTPase is then detected by Western blot using a specific antibody supplied in each kit.

Each GTPase Kit includes GST-fusion protein of the specific binding domain, GTPγS and GDP activators, glutathione agarose resin, lysis-binding-wash buffer, primary antibody, 2X SDS sample buffer, spin columns and collection tubes. Each kit contains sufficient components for 30 pull-down assays. The kits are optimized for Western blot detection with an HRP-conjugated secondary antibody (Part No. 31460, rabbit; Part No. 31430, mouse) using Thermo Scientific SuperSignal™ West Pico Chemiluminescent Substrate (Part No. 34080).

Highlights:

  • High sensitivity and accuracy – optimized reagents, specific antibodies and Western blot procedure ensure accurate controls and quantitative results

  • Convenience – no need to express and purify your own GST-PBD or RBD fusion proteins or use expensive antibody-based immunoprecipitation affinity resins

  • Speed – simultaneous incubation of cell lysate with GST-PBD or RBD and the glutathione resin directly in a high-performance microcentrifuge spin column

  • Ease-of-use – reagents and pull-down conditions have been optimized for immediate success, even for first-time users

  • Efficiency – the microcentrifuge spin columns and collection tubes provide efficient separation of liquid and resin, preventing sample loss

  • Complete – kits include reagents for affinity purification as well as controls, a compatible cell lysis buffer and specific primary antibody for final Western blot detection

GST fusion proteins of active GTPase binding domains:

  • Raf1 Ras-binding domain (Raf1-RBD) binds active Ras
  • Pak1 p21-binding domain (Pak1-PBD) binds active Rac1 and Cdc42
  • Rhotekin Rho-binding domain (Rhotekin-RBD) binds active Rho
  • RalGDS Rap1-binding domain (RalGDS-RBD) binds active Rap1
  • GGA3 protein-binding domain (GGA3-PBD) binds active Arf1 and Arf6
  • Ral binding protein 1 (RalBP1) binds active RalA

Includes:

GST fusion protein of specific binding domain, glutathione agarose resin, GTPγS and GDP (100X), lysis-binding-wash buffer, GTPase-specific primary antibody, SDS-PAGE sample loading buffer (2X), spin columns and collection tubes

Specifications

Content And Storage • GST-Pak1-PBD Fusion Protein, 1 vial 
• Anti-Rac1 Antibody, 5 units, 50 μL 
• Glutathione Agarose Resin, 3 mL 
• GDP (100X), 50 μL 
• GTPyS (100X), 50 μL 
• SDS Sample Buffer (2X), 1.5 mL 
• Lysis/Binding/Wash Buffer (1X), 100 mL 
• Spin Cups, 30 cups 
• Collection Tubes, 90 tubes 

See product manual for storage conditions of individual components.
Description Pierce Active Rac1 Pull-Down and Detection Kit
Assay Protein Quantitation Assay
Conjugate HRP
Detection Method Western Blot, Chemiluminescence
Immunoassay Kit Format Spin Column
Label Type Unlabeled
Substrate Properties Protein-Based Substrate
Substrate Type GTPase Substrate
For Use With (Application) Pull-Down
Kit Contents Spin Cups, 30 cups, SDS Sample Buffer (2X), 1.5mL, Lysis/Binding/Wash Buffer (1X), 100mL, GTPγS (100X), 50μL, GST-Pak1-PBD Fusion Protein, 1 vial, Glutathione Agarose Resin, 3mL, GDP (100X), 50µL, Collection Tubes, 90 tubes, Anti-Rac1 Antibody, 5 units, 50µL
Quantity 30 reactions
Target Kinases (Multiple), Phosphodiesterases, ATPases, GTPases, Phosphatases
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.