missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ 1-Step Transfer Buffer

Catalog No. PI84731X5
Change view
Click to view available options
Quantity:
1 L
5 x 1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI84731X5 5 x 1 L
PI84731 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PI84731X5 Supplier Thermo Scientific™ Supplier No. LSG84731X5

missing translation for 'validMainframeMsg'

Perform rapid semi-dry transfer of 10-300kDa proteins from polyacrylamide gels (SDS-PAGE) to nitrocellulose or PVDF membranes using the Pierce G2 Fast Blotter.

The Thermo Scientific™ Pierce 1-Step Transfer Buffer is designed for rapid semi-dry transfer of 10-300kDa proteins from polyacrylamide gels (SDS-PAGE) to nitrocellulose or PVDF membranes using the Pierce G2 Fast Blotter.

Features of 1-Step Transfer Buffer:

• Fast—high ionic strength formulation provides 5- to 10-minute protein transfer when used with compatible fast semi-dry blotting systems
• Ready to use—supplied as a 1X solution that is stable and ready to use at room temperature
• Optimized—designed for seamless and reliable performance with the Pierce G2 Fast Blotter
• Compatible—effective with other semi-dry transfer devices equipped with a suitable high-current power supply, including the original Pierce Fast Semi-Dry Blotter (Part No. 88217)
• Economical—excellent performance without the special or costly consumables that are required by other fast semi-dry transfer devices

When used with the Pierce G2 Fast Blotter, this buffer provides effective gel-to-membrane transfer of proteins in 5 to 10 minutes with efficiency that is equal to, or better than, conventional Western blotting techniques. Pierce 1-Step Transfer Buffer is also compatible with other protein semi-dry transfer devices, including the original Pierce Fast Semi-Dry Blotter (Part No. 88217), when they are paired with a suitable high-current power supply. Such devices provide constant high current (1.3 to 5.0 amps) to rapidly transfer proteins via the high ionic strength conditions supplied by the transfer buffer.

Requires:
Pierce G2 Fast Blotter (Part No. 62288) or other semi-dry transfer device that is equipped or paired with a capable high-current power supply.

Fast blotting methods require a high-current power supply, such as the Pierce G2 Fast Blotter, and an optimized high ionic strength transfer buffer, such as Pierce 1-Step Transfer Buffer. By increasing the current, excellent transfer efficiency can be achieved in much shorter time compared to conventional methods. Amperage is held at a constant rate based on the surface area of the transfer stack(s) (∼22-23mA/cm2) and voltage is limited to 25V.

Specifications

Product Line Pierce
Product Type Transfer Buffer
Buffer Transfer Buffers
Concentration 1 X
Content And Storage Upon receipt store at room temperature. Product is shipped at ambient temperature.
Gel Type SDS-PAGE Gel (any)
Gel Compatibility SDS-PAGE Gel
Membrane Compatibility Nitrocellulose, PVDF
Quantity 5 x 1 L
I have the Thermo Scientific G2 Fast Blotter (Cat. No. 62288). Has this device been discontinued? Can I get the 1-Step Transfer Buffer recommended for fast transfer with the Thermo Scientific G2 Fast Blotter?

Yes. The Thermo Scientific G2 Fast Blotter has been discontinued. The direct replacement for the Thermo Scientific G2 Fast Blotter is the Thermo Scientific Power Blotter (Cat. No. 22834). The 1-Step Transfer buffer (Cat. No. 84731) is still available.

How many gels can be simultaneously transferred using the Thermo Scientific Power Blotter when used in conjunction with the Thermo Scientific 1-Step Transfer Buffer?

When the Thermo Scientific Power Blotter is used in conjunction with the Thermo Scientific 1-Step Transfer Buffer, one to four mini-sized gels or one to two midi-sized gels can be simultaneously transferred in 5-10 minutes. Please note that gels simultaneously transferred must have the same formulation.

For Research Use Only. Not for use in diagnostic procedures.