missing translation for 'onlineSavingsMsg'
Learn More

Sartorius Picus™ 2 Electronic 8-Channel Pipette PROMO

Catalog No. 01671370PM
Change view
Click to view available options
Volume (Metric):
0.2 to 10 μL
10 to 300 μL
5 to 120 μL
50 to 1200 μL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
01671370PM 50 to 1200 μL
01671371PM 10 to 300 μL
01671372PM 5 to 120 μL
01671373PM 0.2 to 10 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 01671370PM Supplier Sartorius Supplier No. LH747391PM

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Picus™ 2 Electronic 8-Channel Pipette offers reliable, ergonomic pipetting for all users. It has customizable programs, safety features, and connects to the Sartorius app for automatic adjustments and improved workflow, enhancing lab productivity.

  • Ensures reliable and repeatable pipetting results and features an unbeatable ergonomic design that is kind to your hand
  • Picus™ 2 is suited to anyone working in the lab, from a graduate student to an experienced laboratory professional. It is as intuitive to use as a mechanical pipette, but also offers more advanced options for experienced users.
  • Speed up your work: Speed up work by choosing from one of the multiple pipetting modes, which are specifically tailored to ease repetitive work.
  • User-Friendly Electronic Pipetting: Whether you are new to the lab or an expert, the 8-channel Pipette has something to offer every lab professional. It's as easy to start with as a mechanical pipette and offers incredible options for even the most advanced user. You can utilize the build-in plate tracker to increase reliability in microwell plate work.
  • Ready to use: The 8-channel Picus™ Electronic Pipette comes ready to use and partially charged (however, we recommend fully charging it before its first use). For more information on charging, storage, and specifications, consult with Sartorius personnel and additional material available on our site.
  • Reliable and repeatable pipetting results
  • A wide selection of pipetting modes
  • Customizable program
  • Password protection
  • Calibration reminders
  • Sartorius pipetting mobile app

Specifications

Compatible Tips Sartorius Optifit Tips, SafetySpace Tips, Flexibulk Tips
Adjustable Tip Spacing No
Autoclavable Not Autoclavable
Color Gray
Disposable Reusable
For Use With Optifit Tip (10μL), SafetySpace Filter Tip
Grip Type None
Material Polycarbonate ABS
Number of Channels 8
Programmability Yes
Sterility Non-sterile
Type Electronic Pipette
Volume (Metric) 50 to 1200 μL
Fit for Purpose Certified Quality
Show More Show Less