missing translation for 'onlineSavingsMsg'
Learn More

Picrotoxin (Picrotoxinin + Picrotin) 94.0+%, TCI America™

Catalog No. C03751G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C03751G 1 g
C03755G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C03751G Supplier TCI America Supplier No. C03751G

Chemical Identifiers

CAS 124-87-8
Molecular Formula C30H34O13
Molecular Weight (g/mol) 602.589
MDL Number MFCD00074824
InChI Key VJKUPQSHOVKBCO-DMRKGALJSA-N
Synonym picrotoxin
PubChem CID 131846146
SMILES CC(=C)C1C2C3C4(C(C1C(=O)O2)(CC5C4(O5)C(=O)O3)O)C.CC12C3C4C(C(C1(CC5C2(O5)C(=O)O3)O)C(=O)O4)C(C)(C)O

Specifications

Melting Point 200°C
Color White-Yellow
Quantity 1 g
UN Number 2811
Formula Weight 602.59
Percent Purity ≥94.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Picrotoxin (Picrotoxinin + Picrotin)
RTECSNumber TJ9100000
TSCA No