accessibility menu, dialog, popup

UserName

CAS RN 124-87-8

IUPAC Name: (1R,3R,5S,8S,13R,14S)-1-hydroxy-14-(2-hydroxypropan-2-yl)-13-methyl-4,7,10-trioxapentacyclo[6.4.1.1⁹,¹².0³,⁵.0⁵,¹³]tetradecane-6,11-dione; (1R,5S,8S,13R,14R)-1-hydroxy-13-methyl-14-(prop-1-en-2-yl)-4,7,10-trioxapentacyclo[6.4.1.1⁹,¹².0³,⁵.0⁵,¹³]tetradecane-6,11-dione
Synonyms: picrotoxin
Molecular Weight (g/mol): 602.59
Molecular Formula: C30H34O13
InChi Key: VJKUPQSHOVKBCO-ZTYBEOBUSA-N
SMILES: CC(=C)[C@@H]1C2OC(=O)C1[C@]1(O)CC3O[C@]33C(=O)O[C@H]2[C@]13C.CC(C)(O)[C@@H]1C2OC(=O)C1[C@]1(O)C[C@H]3O[C@]33C(=O)O[C@H]2[C@]13C
Molecular Weight (g/mol) 
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (3)
Physical Form 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)