missing translation for 'onlineSavingsMsg'
Learn More

Picrotoxin, 95%

Catalog No. p-4509870
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC131210010 1 g Glass bottle
AC131210050 5 g Glass bottle
AC131210250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC131210010 Supplier Thermo Scientific Chemicals Supplier No. 131210010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 124-87-8 | C30H34O13 | 602.58 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 124-87-8
Molecular Formula C30H34O13
Molecular Weight (g/mol) 602.58
InChI Key VJKUPQSHOVKBCO-DMRKGALJSA-N
Synonym picrotoxin
PubChem CID 131846146
SMILES CC(=C)C1C2C3C4(C(C1C(=O)O2)(CC5C4(O5)C(=O)O3)O)C.CC12C3C4C(C(C1(CC5C2(O5)C(=O)O3)O)C(=O)O4)C(C)(C)O

Specifications

Melting Point 199.0°C to 202.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (Mixture of Picrotoxinin and Picrotin) (HPLC)
Packaging Glass bottle
Quantity 1 g
Merck Index 15,7527
Specific Rotation − 32.00
Solubility Information Solubility in water: 1g/5mL (100°C). Other solubilities: soluble in boiling alcohol (1gr/3mL),dilute acids,and alkalies; sparingly soluble in ether and ccl4,readily soluble in strong ammonia water and in aq.,solns of naoh.,1gr/13.5 mL in 95% ethan
Formula Weight 602.58
Percent Purity 98%
Physical Form Crystals or Powder
Chemical Name or Material Picrotoxin
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Fatal if swallowed.
Precautionary Statement GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
EINECSNumber 204-716-6
Recommended Storage 4°C

RUO – Research Use Only