missing translation for 'onlineSavingsMsg'
Learn More

Picric Acid, Crystal, Spectrum™ Chemical

Catalog No. p-7166117
Change view
Click to view available options
Quantity:
125 g
12 kg
Packaging:
Amber Glass Bottle
Poly Drum
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18612371 125 g Amber Glass Bottle
18612372 12 kg Poly Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18612371 Supplier Spectrum Chemical Mfg Cor Supplier No. P1691125GM
Only null left

Chemical Identifiers

CAS 88-89-1
Molecular Formula C6H3N3O7
Molecular Weight (g/mol) 229.10
InChI Key OXNIZHLAWKMVMX-UHFFFAOYSA-N
IUPAC Name 2,4,6-trinitrophenol
SMILES OC1=C(C=C(C=C1[N+]([O-])=O)[N+]([O-])=O)[N+]([O-])=O

Specifications

Melting Point 121° to 123°C
CAS Max % 1
Grade Reagent
Quantity 125 g
Packaging Amber Glass Bottle

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.