missing translation for 'onlineSavingsMsg'
Learn More

Picloram, 95%

Belongs to the puridine class of the auxinic compounds similar to the hormone Indole-3-acetic acid (IAA) | CAS: 1918-02-1 | C6H3Cl3N2O2 | 241.45 g/mol

Supplier:  Thermo Scientific Chemicals J6670214

 View more versions of this product

Catalog No. AAJ6670214


Only null left
Explore available promotions

Description

Description

Belongs to the puridine class of the auxinic compounds similar to the hormone Indole-3-acetic acid (IAA)

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Picloram
219°C
>110°C (230°F)
C6H3Cl3N2O2
25 g
picloram, 4-amino-3,5,6-trichloropicolinic acid, tordon, borolin, grazon, amdon, 2-pyridinecarboxylic acid, 4-amino-3,5,6-trichloro, k-pin, tordon 10k, tordon 22k
NQQVFXUMIDALNH-UHFFFAOYSA-N
4-amino-3,5,6-trichloropyridine-2-carboxylic acid
15965
241.46
Powder
2-1-1918
421°C
Plastic bottle
MFCD00012101
479075
Soluble in acetone
C1(=C(C(=NC(=C1Cl)Cl)C(=O)O)Cl)N
241.45
CHEBI:34922
95%

Chemical Identifiers

2-1-1918
241.45
NQQVFXUMIDALNH-UHFFFAOYSA-N
15965
4-amino-3,5,6-trichloropyridine-2-carboxylic acid
C6H3Cl3N2O2
MFCD00012101
picloram, 4-amino-3,5,6-trichloropicolinic acid, tordon, borolin, grazon, amdon, 2-pyridinecarboxylic acid, 4-amino-3,5,6-trichloro, k-pin, tordon 10k, tordon 22k
CHEBI:34922
C1(=C(C(=NC(=C1Cl)Cl)C(=O)O)Cl)N
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

P264b-P280i-P305+P351+P338

H319

EINECSNumber : 217-636-1

RTECSNumber : TJ7525000

TSCA : Yes

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only