missing translation for 'onlineSavingsMsg'
Learn More

Picamilon 98.0+%, TCI America™

Catalog No. N09985G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N09985G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N09985G Supplier TCI America Supplier No. N09985G
Only null left

Chemical Identifiers

CAS 34562-97-5
Molecular Formula C10H12N2O3
Molecular Weight (g/mol) 208.217
MDL Number MFCD00186518
InChI Key NAJVRARAUNYNDX-UHFFFAOYSA-N
Synonym N-Nicotinoyl-GABA, N-(3-Carboxypropyl)nicotinamide, 4-(Nicotinamido)butyric Acid, N-Nicotinoyl-gamma-aminobutyric Acid
PubChem CID 60608
IUPAC Name 4-(pyridine-3-carbonylamino)butanoic acid
SMILES C1=CC(=CN=C1)C(=O)NCCCC(=O)O

Specifications

Melting Point 213°C
Color White
Quantity 5 g
Formula Weight 208.22
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Picamilon