accessibility menu, dialog, popup

UserName

CAS RN 34562-97-5

HOONHON

CAS RN 34562-97-5

IUPAC Name: 4-[(pyridin-3-yl)formamido]butanoic acid
Synonyms: picamilon
Molecular Weight (g/mol): 208.22
Molecular Formula: C10H12N2O3
InChi Key: NAJVRARAUNYNDX-UHFFFAOYSA-N
SMILES: OC(=O)CCCNC(=O)C1=CC=CN=C1
Percent Purity 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results