missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PIBF1 Polyclonal Antibody

Sheep Polyclonal Antibody
Supplier: Invitrogen™ PA5149029
Description
Reconstitute at 0.2 mg/mL in sterile PBS.
Mediator of progesterone that by acting on the phospholipase A2 enzyme interferes with arachidonic acid metabolism, induces a Th2 biased immune response, and by controlling NK activity exerts an anti-abortive effect.Chemical Identifiers
| PIBF1 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Polyclonal | |
| Lyophilized | |
| IgG | |
| Human | |
| PIBF1 | |
| C13orf24; Centrosomal protein of 90 kDa; CEP90; PIBF; PIBF1; progesterone immunomodulatory binding factor 1; progesterone-induced blocking factor 1; progesterone-induced-blocking factor 1; RGD1305077; RP11-505F3.1 | |
| PIBF1 | |
| Primary | |
| Antigen affinity chromatography |
| 0.2 mg/mL | |
| Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| Sheep | |
| RUO | |
| PBS with trehalose and no preservative | |
| Q8WXW3 | |
| 10464 | |
| E. coli-derived recombinant human PIBF1, Glu19-Ala419. | |
| Antibody |
Specifications
| PIBF1 | |
| Polyclonal | |
| Unconjugated | |
| PIBF1 | |
| C13orf24; Centrosomal protein of 90 kDa; CEP90; PIBF; PIBF1; progesterone immunomodulatory binding factor 1; progesterone-induced blocking factor 1; progesterone-induced-blocking factor 1; RGD1305077; RP11-505F3.1 | |
| Sheep | |
| Antigen affinity chromatography | |
| RUO | |
| 10464 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.2 mg/mL | |
| PBS with trehalose and no preservative | |
| Q8WXW3 | |
| PIBF1 | |
| E. coli-derived recombinant human PIBF1, Glu19-Ala419. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |