missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PIASy Polyclonal Antibody

Description
A suggested positive control is rat brain tissue lysate. PA5-20954 can be used with blocking peptide PEP-1068.
Functions as an E3-type small ubiquitin-like modifier (SUMO) ligase, stabilizing the interaction between UBE2I and the substrate, and as a SUMO-tethering factor. Plays a crucial role as a transcriptional coregulation in various cellular pathways, including the STAT pathway, the p53/TP53 pathway, the Wnt pathway and the steroid hormone signaling pathway. Involved in gene silencing. Mediates sumoylation of CEBPA, PARK7, HERC2, MYB, TCF4 and RNF168. In Wnt signaling, represses LEF1 and enhances TCF4 transcriptional activities through promoting their sumoylations. Enhances the sumoylation of MTA1 and may participate in its paralog-selective sumoylation. [UniProt].
Specifications
Specifications
| Antigen | PIASy |
| Applications | Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | PIAS4 |
| Gene Accession No. | Q8N2W9, Q9JM05 |
| Gene Alias | e3 sumo- ligase pias4; E3 SUMO-protein ligase PIAS4; FLJ12419; MGC35296; Pias4; Piasg; Pias-gamma; PIASy; protein inhibitor of activated STAT 4; protein inhibitor of activated STAT PIASy; protein inhibitor of activated STAT protein 4; Protein inhibitor of activated STAT protein gamma; protein inhibitor of activated STAT protein PIASy; protein inhibitor of activated STAT, 4; RING-type E3 ubiquitin transferase PIAS4; zinc finger, MIZ-type containing 6; ZMIZ6 |
| Gene Symbols | PIAS4 |
| Show More |