missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phytyl Acetate (cis- and trans- mixture) 88.0+%, TCI America™
Specifications
Specifications
| Color | Yellow |
| Quantity | 25 g |
| Formula Weight | 338.58 |
| Percent Purity | ≥88.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | Phytyl Acetate (cis- and trans- mixture) |
Safety and Handling
| RTECSNumber | MM0675000 |
| TSCA | Yes |