accessibility menu, dialog, popup

UserName

CAS RN 10236-16-5

IUPAC Name: (2E,7R,11R)-3,7,11,15-tetramethylhexadec-2-en-1-yl acetate
Synonyms: (2E,7R,11R)-3,7,11,15-tetramethylhexadec-2-en-1-yl acetate
Molecular Weight (g/mol): 338.58
Molecular Formula: C22H42O2
InChi Key: JIGCTXHIECXYRJ-ILWBRPEASA-N
SMILES: CC(C)CCC[C@@H](C)CCC[C@@H](C)CCC\C(C)=C\COC(C)=O
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results