missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Phytone™ Peptone
GREENER_CHOICE

Catalog No. 292450TF
Change view
Click to view available options
Quantity:
454 g
10 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
292450TF 10 kg
B11906 454 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 292450TF Supplier Gibco™ Supplier No. 292450

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

An animal origin-free (AOF) enzymatic digest of soy.

  • An animal origin-free (AOF) enzymatic digest of soy
  • A nutritious and excellent carbohydrate source due to the naturally high concentrations of vitamins and carbohydrates in soy
  • Customers have successfully used this supplement in CHO-based production of monoclonal antibodies for oncological and rheumatological therapeutics
  • Also used with many different microbial cell types to produce animal health vaccines for swine, such as Clostridial vaccines

Applications of Phytone Peptone include:

  • It is an excellent plant peptone for the cultivation of fungi and fastidious types of bacteria, such as members of the Clostridium and Neisseria genera
  • It has been used in mammalian cell culture applications due to its high carbohydrate content

Specifications

Content And Storage Storage conditions: 2°C to 25°C. Store in cool, dry conditions in well-sealed containers.
Form Powder
Product Type Peptone
For Use With (Application) Production of monoclonal antibodies for oncological and rheumatological therapeutics, microbial cell types to produce animal health vaccines for swine, such as Clostridial vaccines
Product Line Phytone
Quantity 10 kg
Source Animal origin-free, soy