missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phytin, TCI America™
$52.19 - $264.70
Chemical Identifiers
| CAS | 3615-82-5 |
|---|---|
| Molecular Formula | C12H40CaO48P12 |
| Molecular Weight (g/mol) | 1364.167 |
| MDL Number | MFCD00082315 |
| InChI Key | FLIYQIYWLFARGI-UHFFFAOYSA-N |
| Synonym | Calcium Phytate |
| PubChem CID | 131674909 |
| IUPAC Name | calcium;molecular hydrogen;(2,3,4,5,6-pentaphosphonooxycyclohexyl) dihydrogen phosphate |
| SMILES | [HH].[HH].C1(C(C(C(C(C1OP(=O)(O)O)OP(=O)(O)O)OP(=O)(O)O)OP(=O)(O)O)OP(=O)(O)O)OP(=O)(O)O.C1(C(C(C(C(C1OP(=O)(O)O)OP(=O)(O)O)OP(=O)(O)O)OP(=O)(O)O)OP(=O)(O)O)OP(=O)(O)O.[Ca] |
Chemical Identifiers
| 3615-82-5 | |
| 1364.167 | |
| FLIYQIYWLFARGI-UHFFFAOYSA-N | |
| 131674909 | |
| [HH].[HH].C1(C(C(C(C(C1OP(=O)(O)O)OP(=O)(O)O)OP(=O)(O)O)OP(=O)(O)O)OP(=O)(O)O)OP(=O)(O)O.C1(C(C(C(C(C1OP(=O)(O)O)OP(=O)(O)O)OP(=O)(O)O)OP(=O)(O)O)OP(=O)(O)O)OP(=O)(O)O.[Ca] |
| C12H40CaO48P12 | |
| MFCD00082315 | |
| Calcium Phytate | |
| calcium;molecular hydrogen;(2,3,4,5,6-pentaphosphonooxycyclohexyl) dihydrogen phosphate |
Specifications
| 3615-82-5 | |
| C12H40CaO48P12 | |
| 25 g | |
| FLIYQIYWLFARGI-UHFFFAOYSA-N | |
| calcium;molecular hydrogen;(2,3,4,5,6-pentaphosphonooxycyclohexyl) dihydrogen phosphate | |
| 131674909 | |
| Phytin |
| White-Yellow | |
| MFCD00082315 | |
| Calcium Phytate | |
| [HH].[HH].C1(C(C(C(C(C1OP(=O)(O)O)OP(=O)(O)O)OP(=O)(O)O)OP(=O)(O)O)OP(=O)(O)O)OP(=O)(O)O.C1(C(C(C(C(C1OP(=O)(O)O)OP(=O)(O)O)OP(=O)(O)O)OP(=O)(O)O)OP(=O)(O)O)OP(=O)(O)O.[Ca] | |
| 1364.167 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (8)-0488
RTECSNumber : NM7530000
TSCA : Yes