accessibility menu, dialog, popup

UserName

MgCaOPOOOOPOOOOPOOOOPOOOOPOOOOPOOO

CAS RN 3615-82-5

IUPAC Name: calcium magnesium(2+) 2,3,4,5,6-pentakis(phosphonatooxy)cyclohexyl phosphate
Synonyms: phytin
Molecular Weight (g/mol): 712.32
Molecular Formula: C6H6CaMgO24P6
InChi Key: JWJNVODPOMYTLT-UHFFFAOYSA-B
SMILES: [Mg++].[Ca++].[O-]P([O-])(=O)OC1C(OP([O-])([O-])=O)C(OP([O-])([O-])=O)C(OP([O-])([O-])=O)C(OP([O-])([O-])=O)C1OP([O-])([O-])=O
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results