missing translation for 'onlineSavingsMsg'
Learn More

phyproof™ Yohimbine hydrochloride, ≥98% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111001938
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111001938 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111001938 Supplier Merck Emd Millipore Supplier No. PHL82685100MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG

Specifications

CAS 65-19-0
Product Line phyproof
Linear Formula C21H26N2O3 · HCl
MDL Number MFCD00012674
Melting Point 288°C to 290°C
Synonym 17-Hydroxyyohimban-16-carboxylic acid methyl ester hydrochloride
Type Reference Substance
Quantity 100 mg
Formula Weight 390.90
Percent Purity ≥98% (HPLC)

phyproof is a registered trademark of PhytoLab GmbH & Co. KG