missing translation for 'onlineSavingsMsg'
Learn More

phyproof™ Vanillic acid 4-β-D-glucoside, ≥98% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111002170
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111002170 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111002170 Supplier Merck Emd Millipore Supplier No. PHL8985610MG
Only null left

This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG

Specifications

CAS 32142-31-7
Product Line phyproof
Linear Formula C14H18O9
MDL Number MFCD16294851
Synonym 4-Hydroxy-3-methoxybenzoic acid 4-beta-D-glucoside; Vanillic acid 4-beta-D-glucopyranoside
Type Reference Substance
Quantity 10 mg
Formula Weight 330.29
Percent Purity ≥98% (HPLC)

phyproof is a registered trademark of PhytoLab GmbH & Co. KG