missing translation for 'onlineSavingsMsg'
Learn More

phyproof™ trans-Resveratrol, ≥95% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111002044
Change view
Click to view available options
Quantity:
20 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111002044 20 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111002044 Supplier Merck Emd Millipore Supplier No. PHL8953920MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG

Specifications

CAS 501-36-0
Product Line phyproof
Flash Point >200.0°C - closed cup
Linear Formula C14H12O3
MDL Number MFCD00133799
Synonym Resveratrol; 3,4′,5-Trihydroxy-trans-stilbene; 5-[(1 E)-2-(4-Hydroxyphenyl)ethenyl]-1,3-benzenediol
Type Reference Substance
Quantity 20 mg
Formula Weight 228.24
Percent Purity ≥95% (HPLC)
Grade Primary Reference Standard
Show More Show Less

phyproof is a registered trademark of PhytoLab GmbH & Co. KG