missing translation for 'onlineSavingsMsg'
Learn More

phyproof™ trans-Isoferulic acid, ≥95% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111002214
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111002214 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111002214 Supplier Merck Emd Millipore Supplier No. PHL89717100MG

This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG

Specifications

CAS 25522-33-2
Product Line phyproof
Linear Formula C10H10O4
MDL Number MFCD00004391
Synonym trans-3-(3-Hydroxy-4-methoxyphenyl)acrylic acid; trans-3-Hydroxy-4-methoxycinnamic acid; Hesperetic acid
Type Reference Substance
Quantity 100 mg
Formula Weight 194.18
Percent Purity ≥95% (HPLC)
Grade Primary Reference Standard

phyproof is a registered trademark of PhytoLab GmbH & Co. KG