missing translation for 'onlineSavingsMsg'
Learn More

phyproof™ (-)-trans-Caryophyllene, ≥95% (GC), MilliporeSigma™ Supelco™

Catalog No. 111001849
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111001849 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111001849 Supplier Merck Emd Millipore Supplier No. PHL80717100MG

This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG

Specifications

CAS 87-44-5
Product Line phyproof
Density 0.902 g/mL (at 20°C)
Flash Point 96°C
Linear Formula C15H24
MDL Number MFCD00075925
Boiling Point 262°C to 264°C
Synonym beta-Caryophyllene; trans-(1 R,9 S)-8-Methylene-4,11,11-trimethylbicyclo[7.2.0 ]undec-4-ene
Type Reference Substance
Quantity 100 mg
Formula Weight 204.35
Percent Purity ≥95% (GC)
Grade Primary Reference Standard
Show More Show Less

phyproof is a registered trademark of PhytoLab GmbH & Co. KG